Calpeptin, ≥98%
Calpeptin, ≥98%
Calpeptin, ≥98%
Calpeptin, ≥98%
Thermo Scientific Chemicals

Calpeptin, ≥98%

Inhibitor of the calcium-dependent protease calpain and cathepsin L
Have Questions?
Catalog NumberQuantity
J60481.MA
also known as J60481-MA
10 mg
Catalog number J60481.MA
also known as J60481-MA
Price (USD)
238.00
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
238.00
Each
Chemical Identifiers
CAS117591-20-5
IUPAC Namebenzyl N-[(1S)-3-methyl-1-{[(2S)-1-oxohexan-2-yl]carbamoyl}butyl]carbamate
Molecular FormulaC20H30N2O4
InChI KeyPGGUOGKHUUUWAF-ROUUACIJSA-N
SMILESCCCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)OCC1=CC=CC=C1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥97.5%
Calpeptin is used to to inhibit the activities of cathepsin L and calpain-1. Calpeptin blocked the expression of aspartyl protease cathepsin D, amyloid precursor protein, and calparin. It acts as an inhibitor of the Ca2+-dependent protease, calpain. Prevents collagen- and thrombin-induced platelet aggregation, probably by blocking calpain induced phospholipase C and thromboxane synthase activation. Potent cathepsin L inhibitor. It is shown to preferentially inhibit a subset of protein-tyrosine phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C