O-Acetyl-L-carnitine Hydrochloride
O-Acetyl-L-carnitine Hydrochloride
O-Acetyl-L-carnitine Hydrochloride
O-Acetyl-L-carnitine Hydrochloride
Thermo Scientific Chemicals

O-Acetyl-L-carnitine Hydrochloride

Weak cholinergic agonist | CAS: 5080-50-2 | C9H18ClNO4, C9H17NO4·HCl | 239.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J61536.14
also known as J61536-14
Price (USD)
308.65
Special offer
Online exclusive
Ends: 28-Dec-2026
363.00
Save 54.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
308.65
Special offer
Online exclusive
Ends: 28-Dec-2026
363.00
Save 54.35
Each
Chemical Identifiers
CAS5080-50-2
IUPAC Namehydrogen 3-(acetyloxy)-4-(trimethylazaniumyl)butanoate chloride
Molecular FormulaC9H18ClNO4
InChI KeyJATPLOXBFFRHDN-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(=O)OC(CC([O-])=O)C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥96.0 to ≤104.0% (Titration)
FormCrystalline powder or powder
Water Content (Karl Fischer Titration)≤0.5%
O-Acetyl-L-carnitine hydrochloride is used as a cholinergic agonist that stimulates neuronal response to serotonin and acetylcholine. Acetylcarnitine has antinociceptive activity that may be mediated by enhanced activity of muscarinic cholinergic receptors or mGlu2 glutamate receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C