SB 225002
SB 225002
SB 225002
Thermo Scientific Chemicals

SB 225002

A potent, selective and competitive antagonist of the G-protein coupled receptor CXCR2
Have Questions?
Catalog NumberQuantity
J65387.MA
also known as J65387-MA
10 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65387.MA
also known as J65387-MA
Price (USD)
-
Quantity:
10 mg
Request bulk or custom format
Chemical Identifiers
CAS182498-32-4
IUPAC Name3-(2-bromophenyl)-1-(2-hydroxy-4-nitrophenyl)urea
Molecular FormulaC13H10BrN3O4
InChI KeyMQBZVUNNWUIPMK-UHFFFAOYSA-N
SMILESOC1=C(NC(=O)NC2=CC=CC=C2Br)C=CC(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
Assay from Supplier's CofA≥95.0%
FormPowder or crystals or crystalline powder
SB 225002 is a selective non-peptide CXCR2 inhibitor which suppresses IL-8 induced neutrophil chemotaxis and margination. In addition, SB 225002 reduces MPO activity, IL-1β, MIP-2, and keratinocyte-derived chemokine (KC) levels. Further studies suggest that SB 225002 decreases the expression of vascular endothelial growth factor, inducible NOS (NO synthase), and Cox-2 (cyclooxygenase-2 proteins) in colon tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C