HET-0016, 98%
HET-0016, 98%
HET-0016, 98%
Thermo Scientific Chemicals

HET-0016, 98%

Have Questions?
Catalog NumberQuantity
J67168.MA
also known as J67168-MA
10 mg
Catalog number J67168.MA
also known as J67168-MA
Price (USD)
-
Quantity:
10 mg
Request bulk or custom format
Specifications
Assay98%
CAS339068-25-6
DescriptionPale pink
FormSolid
Formula Weight206.29
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C