P32/98, 98%
Thermo Scientific Chemicals

P32/98, 98%

CAS: 136259-20-6 | C9H18N2OS.0.5C4H4O4
Have Questions?
Catalog NumberQuantity
J67268.MA
also known as J67268-MA
10 mg
Catalog number J67268.MA
also known as J67268-MA
Price (USD)
-
Quantity:
10 mg
Request bulk or custom format
Specifications
Assay98%
CAS136259-20-6
DescriptionWhite
FormSolid
Formula Weight260.35
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C