Recombinant Proteinase K, produced in yeast
Thermo Scientific Chemicals

Recombinant Proteinase K, produced in yeast

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J80001.MC
also known as J80001-MC
Price (USD)
117.65
Special offer
Online exclusive
Ends: 28-Dec-2026
138.00
Save 20.35
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
117.65
Special offer
Online exclusive
Ends: 28-Dec-2026
138.00
Save 20.35
Each
Chemical Identifiers
CAS39450-01-6
SpecificationsSpecification SheetSpecification Sheet
CommentA280: Report assay value at 1mg/ml
DNaseNone detected
RNaseNone detected
units/mgdw20 u/mg dry weight
Unit definitionOne Unit liberates one micromole of Folin positive amino acids, measured as tyrosine, at 37?C, pH 7.5, using urea denatured hemoglobin as the substrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.