CCF4-AM
Actual product may vary
CCF4-AM
CCF4-AM, our LiveBLAzer™ substrate, is a lipophilic, esterified form of the CCF4 substrate which allows it to readily enter cells.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K102920mg
K10285mg
Catalog number K1029
Price (USD)
77,400.00
Each
Quantity:
20mg
Price (USD)
77,400.00
Each
CCF4-AM, our LiveBLAzer™ substrate, is a lipophilic, esterified form of the CCF4 substrate which allows it to readily enter cells. Upon entry, cleavage by endogenous cytoplasmic esterases rapidly converts CCF4-AM into its negatively charged form, CCF4, which is retained in the cytosol. CCF4 is a Fluorescence Resonance Energy Transfer (FRET) substrate which consists of a cephalosporin core linking 7-hydroxycoumarin to fluorescein. This product is to be used as a FRET-based substrate for β-lactamase as a sensitive reporter of mammalian gene expression in the GeneBLAzer® technology.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-Permeant
For Use With (Equipment)Flow Cytometer, Microplate Reader
Product LineLiveBLAzer
Quantity20mg
Shipping ConditionRoom Temperature
SubstrateCCF4-AM
Detection MethodFluorescence
FormSolid
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Lactamase Substrate
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).