Product Image
CCF2-FA (free acid)
CCF2-FA (Free Acid) is a fluorescence resonance energy transfer (FRET) substrate that consists of a cephalosporin core linking B7-hydroxycoumarin toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K1034100μg
K10271mg
Catalog number K1034
Price (USD)
862.00
Each
Quantity:
100μg
Price (USD)
862.00
Each
CCF2-FA (Free Acid) is a fluorescence resonance energy transfer (FRET) substrate that consists of a cephalosporin core linking B7-hydroxycoumarin to fluorescein. This product is intended for use as a beta-lactamase substrate in a GeneBLAzer™ cell-based assay.
GeneBLAzer™ cell-based assays typically utilize the membrane-permeant ester forms (CCF2-AM and CCF4-AM) of the negatively charged fluorescent beta-lactamase substrates, CCF2 and CCF4. These lipophilic esters readily enter the cell, where cleavage by endogeneous cytoplasmic esterases rapidly converts them into their negatively charged forms, thereby trapping them in the cytosol.
CCF2-FA is essentially the CCF2 substrate without the esters found in the AM version. CCF2-FA is de-esterified and used in cell lysate applications, bypassing loading across the cell membrane and de-esterification steps. Cell lysates are the preferred method for applications using cells that contain a cell wall. CCF2-FA can also be used as a control to acquire the excitation and emission spectra for CCF2-AM and CCF4-AM.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-Impermeant
Product LineGeneBLAzer
Quantity100μg
Shipping ConditionApproved for shipment on Wet or Dry Ice
SubstrateCCF2-FA
Detection MethodFluorescence
FormPowder
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Lactamase Substrate
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).