NBT/BCIP Reagent Kit
Invitrogen™

NBT/BCIP Reagent Kit

For chromogenic detection of phosphatase.
Have Questions?
Catalog NumberQuantity
N65471 Kit
Catalog number N6547
Price (USD)
470.00
Each
Quantity:
1 Kit
Price (USD)
470.00
Each

The NBT/BCIP Reagent Kit includes 1 g samples of both NBT and BCIP for the chromogenic detection of phosphatase.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Quantity1 Kit
Shipping ConditionRoom Temperature
SubstratePhosphatase Substrate
Detection MethodAbsorbance
Substrate PropertiesChemical Substrate
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (189)

Citations & References
Abstract
Endogenous endothelial cell nitric-oxide synthase modulates apoptosis in cultured breast cancer cells and is transcriptionally regulated by p53.
Authors:Mortensen K,Skouv J,Hougaard DM,Larsson LI
Journal:The Journal of biological chemistry
PubMed ID:10608825
In situ polymerase chain reaction amplification of HIV-1 DNA in brain tissue.
Authors:Strappe PM, Wang TH, McKenzie CA, Lowrie S, Simmonds P, Bell JE
Journal:J Virol Methods
PubMed ID:9562406
A direct in situ polymerase chain reaction (IS-PCR) assay is described for the detection of HIV-1 proviral DNA in formalin fixed paraffin embedded brain tissue. Biotin-16-dUTP is incorporated during the PCR process and microwave pretreatment of tissue sections ensures that no non-specific incorporation into damaged or nicked genomic DNA occurs. ... More
Study of chronic granulomatous disease by a nitroblue tetrazolium densitometric kinetic test: a new research method.
Authors:Iacobini M, Torre A, Palumbo G, Del Principe D, Digilio G, Balducci L
Journal:Clin Chim Acta
PubMed ID:8149637
SPACRCAN, a novel human interphotoreceptor matrix hyaluronan-binding proteoglycan synthesized by photoreceptors and pinealocytes.
Authors:Acharya S, Foletta VC, Lee JW, Rayborn ME, Rodriguez IR, Young WS, Hollyfield JG
Journal:J Biol Chem
PubMed ID:10702256
The interphotoreceptor matrix is a unique extracellular complex occupying the interface between photoreceptors and the retinal pigment epithelium in the fundus of the eye. Because of the putative supportive role in photoreceptor maintenance, it is likely that constituent molecules play key roles in photoreceptor function and may be targets for ... More
Detection of low copy numbers of HPV DNA by fluorescent in situ hybridization combined with confocal microscopy as an alternative to in situ polymerase chain reaction.
Authors:Lizard G, Chignol MC, Souchier C, Roignot P, Chardonnet Y, Schmitt D
Journal:J Virol Methods
PubMed ID:9672129
'In genital lesions infected by human papillomavirus (HPV), histological criteria and HPV DNA typing are of prognostic value. Therefore, non-radioactive methods such as in situ hybridization are used extensively since they preserve the histological organization of the tissue, and allow the detection and characterization of HPV DNA. However, the sensitivity ... More