Fluorescein-CREBtide Substrate

Fluorescein-CREBtide Substrate

A fluorescein-labeled peptide substrate that is based upon the CREB ( cAMP responsive element binding) protein. The peptide substrate isRead more
Have Questions?
Catalog NumberQuantity
PV35081 mg
Catalog number PV3508
Price (USD)
1,216.00
Each
Quantity:
1 mg
Price (USD)
1,216.00
Each
A fluorescein-labeled peptide substrate that is based upon the CREB ( cAMP responsive element binding) protein. The peptide substrate is phosphorylated by a wide range of Ser/Thr kinases.

For use in combination with the LanthaScreen™ Tb-CREB [pSer133] antibody (PV3542) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Amino Acid SequenceFluorescein- KRREILSRRPSYR
Assay EntryBiochemical terbium phospho-specific activity
Label or DyeFITC (Fluorescein)
Molecular Weight (g/mol)2076 Da
Quantity1 mg
Shipping ConditionDry Ice
SubstrateFluorescein-CREBtide
Target EntryAMPK (A2/B1/G1), STK22D, CAMK2B, CHEK1, MAPKAPK2, PASK, PIM2, PRKCN, PRKD2, PRKG2, RPS6KA5, AMPK (A1/B1/G1), AURKA, CAMK1D, CAMK4, CHEK2, MAPKAPK3, PIM1, PKN1, PRKD1, PRKG1, RPS6KA4, CAMK1
Validated KinasesAMPK, AURKA, CAMK1, CAMKID, CAMK2A, CAMK2B, CAMK4, CHEK1, CHEK2, MAPKAPK2, MAPKAPK3, PASK, PIM1, PIM2, PKN1, PRKACA, PRKD1, PRKD2, PRKG1, PRKG2, RPS6KA4, RPS6KA5, STK22D
Detection MethodFluorescence
Substrate PropertiesPeptide-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeSerine/Threonine Kinases
Unit SizeEach
Contents & Storage
1 mg/mL Fluorescein-CREBtide substrate in water. Store at -20°C and protect from light.