Product References
A low-carbohydrate diet induces hepatic insulin resistance and metabolic associated fatty liver disease in mice.
Molecular metabolism
Long F,Bhatti MR,Kellenberger A,Sun W,Modica S,Höring M,Liebisch G,Krieger JP,Wolfrum C,Challa TD
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Neutralization
Wed Mar 01 00:00:00 EST 2023
Lin28B-high breast cancer cells promote immune suppression in the lung pre-metastatic niche via exosomes and support cancer progression.
Nature communications
Qi M,Xia Y,Wu Y,Zhang Z,Wang X,Lu L,Dai C,Song Y,Xu K,Ji W,Zhan L
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Flow Cytometry
Wed Feb 16 00:00:00 EST 2022
Sox2 overexpression alleviates noise-induced hearing loss by inhibiting inflammation-related hair cell apoptosis.
Journal of neuroinflammation
Chen D,Jia G,Zhang Y,Mao H,Zhao L,Li W,Chen Y,Ni Y
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Immunohistochemistry
Mon Feb 28 00:00:00 EST 2022
Sox2 overexpression alleviates noise-induced hearing loss by inhibiting inflammation-related hair cell apoptosis.
Journal of neuroinflammation
Chen D,Jia G,Zhang Y,Mao H,Zhao L,Li W,Chen Y,Ni Y
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Immunohistochemistry
Mon Feb 28 00:00:00 EST 2022
Sox2 overexpression alleviates noise-induced hearing loss by inhibiting inflammation-related hair cell apoptosis.
Journal of neuroinflammation
Chen D,Jia G,Zhang Y,Mao H,Zhao L,Li W,Chen Y,Ni Y
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Immunohistochemistry
Mon Feb 28 00:00:00 EST 2022
Sox2 overexpression alleviates noise-induced hearing loss by inhibiting inflammation-related hair cell apoptosis.
Journal of neuroinflammation
Chen D,Jia G,Zhang Y,Mao H,Zhao L,Li W,Chen Y,Ni Y
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Immunohistochemistry
Mon Feb 28 00:00:00 EST 2022
SARS-CoV-2 Infection of Microglia Elicits Proinflammatory Activation and Apoptotic Cell Death.
Microbiology spectrum
Jeong GU,Lyu J,Kim KD,Chung YC,Yoon GY,Lee S,Hwang I,Shin WH,Ko J,Lee JY,Kwon YC
11-7061-82 was used in Flow cytometry/Cell sorting to report that SARS-CoV-2 can directly infect human microglia, eliciting M1-like proinflammatory responses, followed by cytopathic effects.
Wed Jun 29 00:00:00 EDT 2022
Tumor-derived IL-6 and trans-signaling among tumor, fat, and muscle mediate pancreatic cancer cachexia.
The Journal of experimental medicine
Rupert JE,Narasimhan A,Jengelley DHA,Jiang Y,Liu J,Au E,Silverman LM,Sandusky G,Bonetto A,Cao S,Lu X,O'Connell TM,Liu Y,Koniaris LG,Zimmers TA
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Neutralization
Mon Jun 07 00:00:00 EDT 2021
Restoration of 5-methoxytryptophan protects against atherosclerotic chondrogenesis and calcification in ApoE-/- mice fed high fat diet.
Journal of biomedical science
Lee GL,Liao TL,Wu JY,Wu KK,Kuo CC
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in ELISA
Mon Nov 08 00:00:00 EST 2021
TNF-Like Ligand 1 Aberrance Aggravates Nonalcoholic Steatohepatitis via M1 Macrophage Polarization.
Oxidative medicine and cellular longevity
Luo Y,Guo J,Jia W,Wu M,Yin F,Niu G,Shih DQ,Targan SR,Zhang X
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Flow Cytometry
Fri Mar 04 00:00:00 EST 2022
Murine Model of Sinusitis Infection for Screening Antimicrobial and Immunomodulatory Therapies.
Frontiers in cellular and infection microbiology
Alford MA,Choi KG,Trimble MJ,Masoudi H,Kalsi P,Pletzer D,Hancock REW
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in ELISA
Fri Jul 02 00:00:00 EDT 2021
Betulinic Acid Ameliorates the Severity of Acute Pancreatitis via Inhibition of the NF-κB Signaling Pathway in Mice.
International journal of molecular sciences
Zhou Z,Choi JW,Shin JY,Kim DU,Kweon B,Oh H,Kim YC,Song HJ,Bae GS,Park SJ
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Immunocytochemistry
Sat Jun 26 00:00:00 EDT 2021
Betulinic Acid Ameliorates the Severity of Acute Pancreatitis via Inhibition of the NF-κB Signaling Pathway in Mice.
International journal of molecular sciences
Zhou Z,Choi JW,Shin JY,Kim DU,Kweon B,Oh H,Kim YC,Song HJ,Bae GS,Park SJ
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Immunocytochemistry
Sat Jun 26 00:00:00 EDT 2021
Elevated type I interferon responses potentiate metabolic dysfunction, inflammation, and accelerated aging in mtDNA mutator mice.
Science advances
Lei Y,Guerra Martinez C,Torres-Odio S,Bell SL,Birdwell CE,Bryant JD,Tong CW,Watson RO,West LC,West AP
11-7061 was used in Flow cytometry/Cell sorting to further advance our knowledge of how mitochondrial dysfunction shapes innate immune responses and provide a framework for understanding mitochondria-driven immunopathology in POLG-related disorders and aging.
Sat May 01 00:00:00 EDT 2021
Spatial proteomics revealed a CX3CL1-dependent crosstalk between the urothelium and relocated macrophages through IL-6 during an acute bacterial infection in the urinary bladder.
Mucosal immunology
Bottek J,Soun C,Lill JK,Dixit A,Thiebes S,Beerlage AL,Horstmann M,Urbanek A,Heuer H,Uszkoreit J,Eisenacher M,Bracht T,Sitek B,Hoffmann F,Vijitha N,von Eggeling F,Engel DR
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in ELISA
Wed Jul 01 00:00:00 EDT 2020
Spatial proteomics revealed a CX3CL1-dependent crosstalk between the urothelium and relocated macrophages through IL-6 during an acute bacterial infection in the urinary bladder.
Mucosal immunology
Bottek J,Soun C,Lill JK,Dixit A,Thiebes S,Beerlage AL,Horstmann M,Urbanek A,Heuer H,Uszkoreit J,Eisenacher M,Bracht T,Sitek B,Hoffmann F,Vijitha N,von Eggeling F,Engel DR
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in ELISA
Wed Jul 01 00:00:00 EDT 2020
Regulating colonic dendritic cells by commensal glycosylated large surface layer protein A to sustain gut homeostasis against pathogenic inflammation.
Mucosal immunology
Ge Y,Gong M,Zadeh M,Li J,Abbott JR,Li W,Morel L,Sonon R,Supekar NT,Azadi P,Wang Y,Jones DP,Li S,Mohamadzadeh M
11-7061 was used in Flow cytometry/Cell sorting to study the molecular mechanisms responsible for regulating gut homeostasis.
Wed Jan 01 00:00:00 EST 2020
Senolytics prevent mt-DNA-induced inflammation and promote the survival of aged organs following transplantation.
Nature communications
Iske J,Seyda M,Heinbokel T,Maenosono R,Minami K,Nian Y,Quante M,Falk CS,Azuma H,Martin F,Passos JF,Niemann CU,Tchkonia T,Kirkland JL,Elkhal A,Tullius SG
11-7061 was used in Flow cytometry/Cell sorting to show that cell-free mitochondrial DNA (cf-mt-DNA) released by senescent cells accumulates with aging and augments immunogenicity.
Thu Aug 27 00:00:00 EDT 2020
Peli1 controls the survival of dopaminergic neurons through modulating microglia-mediated neuroinflammation.
Scientific reports
Dai D,Yuan J,Wang Y,Xu J,Mao C,Xiao Y
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in ELISA
Wed May 29 00:00:00 EDT 2019
Vanadium Derivative Exposure Promotes Functional Alterations of VSMCs and Consequent Atherosclerosis via ROS/p38/NF-κB-Mediated IL-6 Production.
International journal of molecular sciences
Yeh CC,Wu JY,Lee GL,Wen HT,Lin P,Kuo CC
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in ELISA
Wed Dec 04 00:00:00 EST 2019
1,25-Dihydroxyvitamin D suppresses M1 macrophages and promotes M2 differentiation at bone injury sites.
JCI insight
Wasnik S,Rundle CH,Baylink DJ,Yazdi MS,Carreon EE,Xu Y,Qin X,Lau KW,Tang X
11-7061 was used in Flow cytometry/Cell sorting to define the role of M1 macrophages at bone injury sites via the function of 1,25-Dihydroxyvitamin D in suppressing M1 but promoting M2 differentiation.
Thu Sep 06 00:00:00 EDT 2018
Hepatic inflammation caused by dysregulated bile acid synthesis is reversible by butyrate supplementation.
The Journal of pathology
Sheng L,Jena PK,Hu Y,Liu HX,Nagar N,Kalanetra KM,French SW,French SW,Mills DA,Wan YY
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Western Blot
Fri Dec 01 00:00:00 EST 2017
Depletion of microglia exacerbates postischemic inflammation and brain injury.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Jin WN,Shi SX,Li Z,Li M,Wood K,Gonzales RJ,Liu Q
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Tumor-associated macrophages promote neuroblastoma via STAT3 phosphorylation and up-regulation of c-MYC.
Oncotarget
Hadjidaniel MD,Muthugounder S,Hung LT,Sheard MA,Shirinbak S,Chan RY,Nakata R,Borriello L,Malvar J,Kennedy RJ,Iwakura H,Akamizu T,Sposto R,Shimada H,DeClerck YA,Asgharzadeh S
Tue Oct 31 00:00:00 EDT 2017
Sprouty1 induces a senescence-associated secretory phenotype by regulating NFκB activity: implications for tumorigenesis.
Cell death and differentiation
Macià A,Vaquero M,Gou-Fàbregas M,Castelblanco E,Valdivielso JM,Anerillas C,Mauricio D,Matias-Guiu X,Ribera J,Encinas M
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Neutralization
Sat Feb 01 00:00:00 EST 2014
Live single cell functional phenotyping in droplet nano-liter reactors.
Scientific reports
Konry T,Golberg A,Yarmush M
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Immunofluorescence
Mon Nov 11 00:00:00 EST 2013
Osteoclast activated FoxP3+ CD8+ T-cells suppress bone resorption in vitro.
PloS one
Buchwald ZS,Kiesel JR,DiPaolo R,Pagadala MS,Aurora R
11-7061 was used in Flow cytometry/Cell sorting to demonstrate the effects of activated CD8 T cell-produced cytokines on osteoclast activity.
Thu Nov 08 00:00:00 EST 2012
CD8 regulates T regulatory cell production of IL-6 and maintains their suppressive phenotype in allergic lung disease.
Journal of immunology (Baltimore, Md. : 1950)
Joetham A,Okamoto M,Takeda K,Schedel M,Ohnishi H,Dakhama A,Gelfand EW
11-7061 was used in Flow cytometry/Cell sorting to identify the critical role of CD8-T regulatory cell interactions in regulating the suppressive phenotype of nTregs through control of IL-6 production.
Sat Jan 01 00:00:00 EST 2011
IL-6 produced by immune complex-activated follicular dendritic cells promotes germinal center reactions, IgG responses and somatic hypermutation.
International immunology
Wu Y,El Shikh ME,El Sayed RM,Best AM,Szakal AK,Tew JG
Published figure using IL-6 monoclonal antibody (Product # 11-7061-82) in Neutralization
Mon Jun 01 00:00:00 EDT 2009