Product References
R274X-mutated Phf6 increased the self-renewal and skewed T cell differentiation of hematopoietic stem cells.
iScience
Lan Y,Yuan S,Guo T,Hou S,Zhao F,Yang W,Cao Y,Chu Y,Jiang E,Yuan W,Wang X
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Fri Jun 16 00:00:00 EDT 2023
R274X-mutated Phf6 increased the self-renewal and skewed T cell differentiation of hematopoietic stem cells.
iScience
Lan Y,Yuan S,Guo T,Hou S,Zhao F,Yang W,Cao Y,Chu Y,Jiang E,Yuan W,Wang X
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Fri Jun 16 00:00:00 EDT 2023
LINC01123 promotes immune escape by sponging miR-214-3p to regulate B7-H3 in head and neck squamous-cell carcinoma.
Cell death & disease
Li H,Yang Z,Yang X,Zhang F,Wang J,Wu Z,Wanyan C,Meng Q,Gao W,Yang X,Wei J
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Thu Feb 03 00:00:00 EST 2022
PRDM1/BLIMP1 induces cancer immune evasion by modulating the USP22-SPI1-PD-L1 axis in hepatocellular carcinoma cells.
Nature communications
Li Q,Zhang L,You W,Xu J,Dai J,Hua D,Zhang R,Yao F,Zhou S,Huang W,Dai Y,Zhang Y,Baheti T,Qian X,Pu L,Xu J,Xia Y,Zhang C,Tang J,Wang X
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Mon Dec 12 00:00:00 EST 2022
Transcriptional correlates of malaria in RTS,S/AS01-vaccinated African children: a matched case-control study.
eLife
Moncunill G,Carnes J,Chad Young W,Carpp L,De Rosa S,Campo JJ,Nhabomba A,Mpina M,Jairoce C,Finak G,Haas P,Muriel C,Van P,Sanz H,Dutta S,Mordmüller B,Agnandji ST,Díez-Padrisa N,Williams NA,Aponte JJ,Valim C,Neafsey DE,Daubenberger C,McElrath MJ,Dobaño C,Stuart K,Gottardo R
11-7349-82 was used in Flow cytometry/Cell sorting to understand RTS,S/AS01-induced immune responses associated with vaccine protection.
Fri Jan 21 00:00:00 EST 2022
Primary exposure to SARS-CoV-2 variants elicits convergent epitope specificities, immunoglobulin V gene usage and public B cell clones.
Nature communications
Lima NS,Musayev M,Johnston TS,Wagner DA,Henry AR,Wang L,Yang ES,Zhang Y,Birungi K,Black WP,O'Dell S,Schmidt SD,Moon D,Lorang CG,Zhao B,Chen M,Boswell KL,Roberts-Torres J,Davis RL,Peyton L,Narpala SR,O'Connell S,Serebryannyy L,Wang J,Schrager A,Talana CA,Shimberg G,Leung K,Shi W,Khashab R,Biber A,Zilberman T,Rhein J,Vetter S,Ahmed A,Novik L,Widge A,Gordon I,Guech M,Teng IT,Phung E,Ruckwardt TJ,Pegu A,Misasi J,Doria-Rose NA,Gaudinski M,Koup RA,Kwong PD,McDermott AB,Amit S,Schacker TW,Levy I,Mascola JR,Sullivan NJ,Schramm CA,Douek DC
11-7349-82 was used in Immunocytochemistry to develop a method to obtain immunoglobulin sequences with concurrent rapid production and functional assessment of monoclonal antibodies from hundreds of single B cells sorted by flow cytometry.
Wed Dec 14 00:00:00 EST 2022
Baricitinib treatment resolves lower-airway macrophage inflammation and neutrophil recruitment in SARS-CoV-2-infected rhesus macaques.
Cell
Hoang TN,Pino M,Boddapati AK,Viox EG,Starke CE,Upadhyay AA,Gumber S,Nekorchuk M,Busman-Sahay K,Strongin Z,Harper JL,Tharp GK,Pellegrini KL,Kirejczyk S,Zandi K,Tao S,Horton TR,Beagle EN,Mahar EA,Lee MYH,Cohen J,Jean SM,Wood JS,Connor-Stroud F,Stammen RL,Delmas OM,Wang S,Cooney KA,Sayegh MN,Wang L,Filev PD,Weiskopf D,Silvestri G,Waggoner J,Piantadosi A,Kasturi SP,Al-Shakhshir H,Ribeiro SP,Sekaly RP,Levit RD,Estes JD,Vanderford TH,Schinazi RF,Bosinger SE,Paiardini M
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Thu Jan 21 00:00:00 EST 2021
Membrane lymphotoxin-α2β is a novel tumor necrosis factor (TNF) receptor 2 (TNFR2) agonist.
Cell death & disease
Kucka K,Lang I,Zhang T,Siegmund D,Medler J,Wajant H
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Tue Apr 06 00:00:00 EDT 2021
NKD2 mediates stimulation-dependent ORAI1 trafficking to augment Ca2+ entry in T cells.
Cell reports
Wu B,Woo JS,Vila P,Jew M,Leung J,Sun Z,Srikanth S,Gwack Y
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Tue Aug 24 00:00:00 EDT 2021
Development of an Inflammatory CD14+ Dendritic Cell Subset in Humanized Mice.
Frontiers in immunology
Iwabuchi R,Ide K,Terahara K,Wagatsuma R,Iwaki R,Matsunaga H,Tsunetsugu-Yokota Y,Takeyama H,Takahashi Y
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Mon Sep 27 00:00:00 EDT 2021
Rejuvenating Effector/Exhausted CAR T Cells to Stem Cell Memory-Like CAR T Cells By Resting Them in the Presence of CXCL12 and the NOTCH Ligand.
Cancer research communications
Ando M,Kondo T,Tomisato W,Ito M,Shichino S,Srirat T,Mise-Omata S,Nakagawara K,Yoshimura A
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
Inhibition of Cholesterol Esterification Enzyme Enhances the Potency of Human Chimeric Antigen Receptor T Cells against Pancreatic Carcinoma.
Molecular therapy oncolytics
Zhao L,Liu Y,Zhao F,Jin Y,Feng J,Geng R,Sun J,Kang L,Yu L,Wei Y
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Fri Mar 27 00:00:00 EDT 2020
Abrogation of HLA surface expression using CRISPR/Cas9 genome editing: a step toward universal T cell therapy.
Scientific reports
Lee J,Sheen JH,Lim O,Lee Y,Ryu J,Shin D,Kim YY,Kim M
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Tue Oct 20 00:00:00 EDT 2020
Ghrelin ameliorates tumor-induced adipose tissue atrophy and inflammation via Ghrelin receptor-dependent and -independent pathways.
Oncotarget
Liu H,Luo J,Guillory B,Chen JA,Zang P,Yoeli JK,Hernandez Y,Lee II,Anderson B,Storie M,Tewnion A,Garcia JM
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Immunohistochemistry
Tue Sep 01 00:00:00 EDT 2020
Ghrelin ameliorates tumor-induced adipose tissue atrophy and inflammation via Ghrelin receptor-dependent and -independent pathways.
Oncotarget
Liu H,Luo J,Guillory B,Chen JA,Zang P,Yoeli JK,Hernandez Y,Lee II,Anderson B,Storie M,Tewnion A,Garcia JM
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Immunohistochemistry
Tue Sep 01 00:00:00 EDT 2020
Mucosal-associated invariant T (MAIT) cells mediate protective host responses in sepsis.
eLife
Trivedi S,Labuz D,Anderson CP,Araujo CV,Blair A,Middleton EA,Jensen O,Tran A,Mulvey MA,Campbell RA,Hale JS,Rondina MT,Leung DT
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Mon Nov 09 00:00:00 EST 2020
CAIX Regulates GBM Motility and TAM Adhesion and Polarization through EGFR/STAT3 under Hypoxic Conditions.
International journal of molecular sciences
Huang BR,Liu YS,Lai SW,Lin HJ,Shen CK,Yang LY,Lu DY
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Fri Aug 14 00:00:00 EDT 2020
Siglec-9 Regulates an Effector Memory CD8+ T-cell Subset That Congregates in the Melanoma Tumor Microenvironment.
Cancer immunology research
Haas Q,Boligan KF,Jandus C,Schneider C,Simillion C,Stanczak MA,Haubitz M,Seyed Jafari SM,Zippelius A,Baerlocher GM,Läubli H,Hunger RE,Romero P,Simon HU,von Gunten S
11-7349 was used in Flow cytometry/Cell sorting to demonstrate cytotoxic CD8+ T cells in melanoma intratumoral blood express Siglec-9.
Wed May 01 00:00:00 EDT 2019
Exhaustion of the CD8+ T Cell Compartment in Patients with Mutations in Phosphoinositide 3-Kinase Delta.
Frontiers in immunology
Wentink MWJ,Mueller YM,Dalm VASH,Driessen GJ,van Hagen PM,van Montfrans JM,van der Burg M,Katsikis PD
11-7349 was used in Flow cytometry/Cell sorting to study exhaustion of the CD8+ T cell compartment in activated phosphoinositide 3-kinase delta syndrome patients and compare them with healthy controls and HIV patients.
Mon Jun 24 00:00:00 EDT 2019
Heterogeneous Tumor-Immune Microenvironments among Differentially Growing Metastases in an Ovarian Cancer Patient.
Cell
Jiménez-Sánchez A,Memon D,Pourpe S,Veeraraghavan H,Li Y,Vargas HA,Gill MB,Park KJ,Zivanovic O,Konner J,Ricca J,Zamarin D,Walther T,Aghajanian C,Wolchok JD,Sala E,Merghoub T,Snyder A,Miller ML
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Thu Aug 24 00:00:00 EDT 2017
The distribution and function of human memory T cell subsets in lung cancer.
Immunologic research
Sheng SY,Gu Y,Lu CG,Zou JY,Hong H,Wang R
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Application of a whole blood mycobacterial growth inhibition assay to study immunity against Mycobacterium tuberculosis in a high tuberculosis burden population.
PloS one
Baguma R,Penn-Nicholson A,Smit E,Erasmus M,Day J,Makhethe L,de Kock M,Hughes EJ,van Rooyen M,Pienaar B,Stone L,Hanekom W,Brennan MJ,Wallis RS,Hatherill M,Scriba TJ
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Wed Oct 18 00:00:00 EDT 2017
High pathogen burden in childhood promotes the development of unconventional innate-like CD8+ T cells.
JCI insight
Falanga YT,Frascoli M,Kaymaz Y,Forconi C,Ong'echa JM,Bailey JA,Berg LJ,Moormann AM
11-7349 was used in Flow cytometry/Cell sorting to suggest that the unconventional CD8dim T cells arise in a very particular immunological context and may provide a deeper understanding of the heterogeneity in human immune responses.
Thu Aug 03 00:00:00 EDT 2017
Compartment-specific immunity in the human gut: properties and functions of dendritic cells in the colon versus the ileum.
Gut
Mann ER,Bernardo D,English NR,Landy J,Al-Hassi HO,Peake ST,Man R,Elliott TR,Spranger H,Lee GH,Parian A,Brant SR,Lazarev M,Hart AL,Li X,Knight SC
11-7349 was used in Flow cytometry/Cell sorting to investigate and distinguish properties of dendritic cells in the colon and the ileum.
Mon Feb 01 00:00:00 EST 2016
Human Infant Memory B Cell and CD4+ T Cell Responses to HibMenCY-TT Glyco-Conjugate Vaccine.
PloS one
Fuery A,Richmond PC,Currie AJ
11-7349 was used in Flow cytometry/Cell sorting to demonstrate that Men C-specific memory B cells present pre-boost are influential in secondary IgG responses to HibMenCY-TT vaccine.
Thu Apr 28 00:00:00 EDT 2016
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Neutralization
Tue Aug 12 00:00:00 EDT 2014
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Neutralization
Tue Aug 12 00:00:00 EDT 2014
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Neutralization
Tue Aug 12 00:00:00 EDT 2014
Targeting aPKC disables oncogenic signaling by both the EGFR and the proinflammatory cytokine TNFα in glioblastoma.
Science signaling
Kusne Y,Carrera-Silva EA,Perry AS,Rushing EJ,Mandell EK,Dietrich JD,Errasti AE,Gibbs D,Berens ME,Loftus JC,Hulme C,Yang W,Lu Z,Aldape K,Sanai N,Rothlin CV,Ghosh S
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Neutralization
Tue Aug 12 00:00:00 EDT 2014
Attenuation of natural killer cell functions by capsaicin through a direct and TRPV1-independent mechanism.
Carcinogenesis
Kim HS,Kwon HJ,Kim GE,Cho MH,Yoon SY,Davies AJ,Oh SB,Lee H,Cho YK,Joo CH,Kwon SW,Kim SC,Kim YK
11-7349 was used in Flow cytometry/Cell sorting to demonstrate that capsaicin inhibits NK cell-mediated cytotoxicity and cytokine production (interferon-γ and tumor necrosis factor-α).
Tue Jul 01 00:00:00 EDT 2014
Attenuation of natural killer cell functions by capsaicin through a direct and TRPV1-independent mechanism.
Carcinogenesis
Kim HS,Kwon HJ,Kim GE,Cho MH,Yoon SY,Davies AJ,Oh SB,Lee H,Cho YK,Joo CH,Kwon SW,Kim SC,Kim YK
11-7349 was used in Flow cytometry/Cell sorting to demonstrate that capsaicin inhibits NK cell-mediated cytotoxicity and cytokine production (interferon-γ and tumor necrosis factor-α).
Tue Jul 01 00:00:00 EDT 2014
Attenuation of natural killer cell functions by capsaicin through a direct and TRPV1-independent mechanism.
Carcinogenesis
Kim HS,Kwon HJ,Kim GE,Cho MH,Yoon SY,Davies AJ,Oh SB,Lee H,Cho YK,Joo CH,Kwon SW,Kim SC,Kim YK
11-7349 was used in Flow cytometry/Cell sorting to demonstrate that capsaicin inhibits NK cell-mediated cytotoxicity and cytokine production (interferon-γ and tumor necrosis factor-α).
Tue Jul 01 00:00:00 EDT 2014
Attenuation of natural killer cell functions by capsaicin through a direct and TRPV1-independent mechanism.
Carcinogenesis
Kim HS,Kwon HJ,Kim GE,Cho MH,Yoon SY,Davies AJ,Oh SB,Lee H,Cho YK,Joo CH,Kwon SW,Kim SC,Kim YK
11-7349 was used in Flow cytometry/Cell sorting to demonstrate that capsaicin inhibits NK cell-mediated cytotoxicity and cytokine production (interferon-γ and tumor necrosis factor-α).
Tue Jul 01 00:00:00 EDT 2014
Cell-mediated immune responses to respiratory syncytial virus infection: magnitude, kinetics, and correlates with morbidity and age.
Human vaccines & immunotherapeutics
Geevarghese B,Weinberg A
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Mon Apr 25 00:00:00 EDT 2016
Memory T cells in latent Mycobacterium tuberculosis infection are directed against three antigenic islands and largely contained in a CXCR3+CCR6+ Th1 subset.
PLoS pathogens
Lindestam Arlehamn CS,Gerasimova A,Mele F,Henderson R,Swann J,Greenbaum JA,Kim Y,Sidney J,James EA,Taplitz R,McKinney DM,Kwok WW,Grey H,Sallusto F,Peters B,Sette A
11-7349 was used in Flow cytometry/Cell sorting to further define the immunological footprint of Mycobacterium tuberculosis CD4 T cell recognition.
Tue Jan 01 00:00:00 EST 2013
Functional OCT4-specific CD4+ and CD8+ T cells in healthy controls and ovarian cancer patients.
Oncoimmunology
Di J,Massuger LF,Duiveman-de Boer T,Zusterzeel PL,Figdor CG,Torensma R
Published figure using TNF alpha monoclonal antibody (Product # 11-7349-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2013
Expansion of polyfunctional HIV-specific T cells upon stimulation with mRNA electroporated dendritic cells in the presence of immunomodulatory drugs.
Journal of virology
De Keersmaecker B,Allard SD,Lacor P,Schots R,Thielemans K,Aerts JL
11-7349 was used in Immunofluorescence to provide new information about the effects of IMiDs on antigen-specific T cells and suggest that these drugs increase the efficacy of immune therapies for infectious diseases and cancer.
Sat Sep 01 00:00:00 EDT 2012
Durable mucosal simian immunodeficiency virus-specific effector memory T lymphocyte responses elicited by recombinant adenovirus vectors in rhesus monkeys.
Journal of virology
Li H,Liu J,Carville A,Mansfield KG,Lynch D,Barouch DH
11-7349 was used in Flow cytometry/Cell sorting to demonstrate that nonreplicating rAd vectors induce durable and widely distributed effector memory mucosal T lymphocyte responses.
Tue Nov 01 00:00:00 EDT 2011