Product References
Age-related trabecular bone loss is associated with a decline in serum Galectin-1 level.
BMC musculoskeletal disorders
Xu W,Ni C,Wang Y,Zheng G,Zhang J,Xu Y
12-0731 was used in Flow cytometry/Cell sorting to illustrate the relationship of serum Gal-1 level with bone loss.
Tue Apr 27 00:00:00 EDT 2021
Age-related trabecular bone loss is associated with a decline in serum Galectin-1 level.
BMC musculoskeletal disorders
Xu W,Ni C,Wang Y,Zheng G,Zhang J,Xu Y
12-0731 was used in Flow cytometry/Cell sorting to illustrate the relationship of serum Gal-1 level with bone loss.
Tue Apr 27 00:00:00 EDT 2021
Transplantation of bone marrow-derived mesenchymal stem cells with silencing of microRNA-138 relieves pelvic organ prolapse through the FBLN5/IL-1β/elastin pathway.
Aging
Zhao B,Sun Q,Fan Y,Hu X,Li L,Wang J,Cui S
Published figure using CD73 monoclonal antibody (Product # 12-0731-82) in Flow Cytometry
Sat Jan 16 00:00:00 EST 2021
Adipose-derived stem cells therapy effectively attenuates PM2.5-induced lung injury.
Stem cell research & therapy
Gao J,Yuan J,Liu Q,Wang Y,Wang H,Chen Y,Ding W,Ji G,Lu Z
12-0731-82 was used in Flow Cytometry to suggest that ADSC transplantation may be a potential therapeutic approach for severe air pollution-associated diseases.
Sat Jun 19 00:00:00 EDT 2021
Negative feedback control of neuronal activity by microglia.
Nature
Badimon A,Strasburger HJ,Ayata P,Chen X,Nair A,Ikegami A,Hwang P,Chan AT,Graves SM,Uweru JO,Ledderose C,Kutlu MG,Wheeler MA,Kahan A,Ishikawa M,Wang YC,Loh YE,Jiang JX,Surmeier DJ,Robson SC,Junger WG,Sebra R,Calipari ES,Kenny PJ,Eyo UB,Colonna M,Quintana FJ,Wake H,Gradinaru V,Schaefer A
12-0731 was used in Flow cytometry/Cell sorting to show that microglia are also critical modulators of neuronal activity and associated behavioural responses in mice.
Thu Oct 01 00:00:00 EDT 2020
Negative feedback control of neuronal activity by microglia.
Nature
Badimon A,Strasburger HJ,Ayata P,Chen X,Nair A,Ikegami A,Hwang P,Chan AT,Graves SM,Uweru JO,Ledderose C,Kutlu MG,Wheeler MA,Kahan A,Ishikawa M,Wang YC,Loh YE,Jiang JX,Surmeier DJ,Robson SC,Junger WG,Sebra R,Calipari ES,Kenny PJ,Eyo UB,Colonna M,Quintana FJ,Wake H,Gradinaru V,Schaefer A
12-0731 was used in Flow cytometry/Cell sorting to show that microglia are also critical modulators of neuronal activity and associated behavioural responses in mice.
Thu Oct 01 00:00:00 EDT 2020
Targeting the ion channel TRPM7 promotes the thymic development of regulatory T cells by promoting IL-2 signaling.
Science signaling
Mendu SK,Stremska ME,Schappe MS,Moser EK,Krupa JK,Rogers JS,Stipes EJ,Parker CA,Braciale TJ,Perry JSA,Desai BN
12-0731-82 was used in Flow Cytometry to demonstrate that inhibition of the dual ion channel and enzyme, TRPM7, increases thymic development of regulatory T cells in mice.
Tue Dec 08 00:00:00 EST 2020
Sema3A and HIF1α co-overexpressed iPSC-MSCs/HA scaffold facilitates the repair of calvarial defect in a mouse model.
Journal of cellular physiology
Li J,Wang T,Li C,Wang Z,Wang P,Zheng L
12-0731-82 was used in Flow cytometry/Cell sorting to show that Sema3A and HIF1α co-overexpression reversed the reduced cell proliferation that reduced by Sema3A overexpression alone.
Thu Oct 01 00:00:00 EDT 2020
Sema3A and HIF1α co-overexpressed iPSC-MSCs/HA scaffold facilitates the repair of calvarial defect in a mouse model.
Journal of cellular physiology
Li J,Wang T,Li C,Wang Z,Wang P,Zheng L
12-0731-82 was used in Flow cytometry/Cell sorting to show that Sema3A and HIF1α co-overexpression reversed the reduced cell proliferation that reduced by Sema3A overexpression alone.
Thu Oct 01 00:00:00 EDT 2020
Effect of mesenchymal stem cell-derived exosomes on the induction of mouse tolerogenic dendritic cells.
Journal of cellular physiology
Shahir M,Mahmoud Hashemi S,Asadirad A,Varahram M,Kazempour-Dizaji M,Folkerts G,Garssen J,Adcock I,Mortaz E
12-0731 was used in Flow cytometry/Cell sorting to investigate the effects of mesenchymal stem cell-derived exosomes and to understand their role in modulating the mouse tolergenic dendritic cell immune responses.
Thu Oct 01 00:00:00 EDT 2020
Extracellular adenosine enhances the ability of PMNs to kill Streptococcus pneumoniae by inhibiting IL-10 production.
Journal of leukocyte biology
Siwapornchai N,Lee JN,Tchalla EYI,Bhalla M,Yeoh JH,Roggensack SE,Leong JM,Bou Ghanem EN
12-0731-82 was used in Flow Cytometry to explore why the antimicrobial efficacy of polymorphonuclear leukocytes declines over the course of infection.
Tue Sep 01 00:00:00 EDT 2020
CD73 Promotes Glioblastoma Pathogenesis and Enhances Its Chemoresistance via A2B Adenosine Receptor Signaling.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Yan A,Joachims ML,Thompson LF,Miller AD,Canoll PD,Bynoe MS
Published figure using CD73 monoclonal antibody (Product # 12-0731-82) in Flow Cytometry
Wed May 29 00:00:00 EDT 2019
Transcriptional Signature Derived from Murine Tumor-Associated Macrophages Correlates with Poor Outcome in Breast Cancer Patients.
Cell reports
Tuit S,Salvagno C,Kapellos TS,Hau CS,Seep L,Oestreich M,Klee K,de Visser KE,Ulas T,Schultze JL
12-0731 was used in Flow cytometry/Cell sorting to show that a transgenic mouse tumor model can yield a TAM signature relevant for human breast cancer outcome prognosis and provide a generalizable strategy for determining and applying immune cell signatures provided the murine model reflects the human disease.
Tue Oct 29 00:00:00 EDT 2019
Dysregulation of ectonucleotidase-mediated extracellular adenosine during postmenopausal bone loss.
Science advances
Shih YV,Liu M,Kwon SK,Iida M,Gong Y,Sangaj N,Varghese S
12-0731-82 was used in Flow cytometry/Cell sorting to demonstrate that CD39/CD73 expression and extracellular adenosine levels in the bone marrow are substantially decreased in animals with osteoporotic bone loss.
Thu Aug 01 00:00:00 EDT 2019
PDGF is a potent initiator of bone formation in a tissue engineered model of pathological ossification.
Journal of tissue engineering and regenerative medicine
Davies OG,Grover LM,Lewis MP,Liu Y
Published figure using CD73 monoclonal antibody (Product # 12-0731-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Rap1 deficiency-provoked paracrine dysfunction impairs immunosuppressive potency of mesenchymal stem cells in allograft rejection of heart transplantation.
Cell death & disease
Ding Y,Liang X,Zhang Y,Yi L,Shum HC,Chen Q,Chan BP,Fan H,Liu Z,Tergaonkar V,Qi Z,Tse HF,Lian Q
12-0731 was used in Flow cytometry/Cell sorting to indicate that Rap1 is essential to maintain the immunomodulatory function of MSCs.
Wed Mar 07 00:00:00 EST 2018
A population of hematopoietic stem cells derives from GATA4-expressing progenitors located in the placenta and lateral mesoderm of mice.
Haematologica
Cañete A,Carmona R,Ariza L,Sánchez MJ,Rojas A,Muñoz-Chápuli R
12073182 was used in flow cytometry to utilize a G2-Gata4Cre;R26REYFP mouse line to track the developmental fate of the G2-Gata4 cell lineage
Sat Apr 01 00:00:00 EDT 2017
Calcium-mediated shaping of naive CD4 T-cell phenotype and function.
eLife
Guichard V,Bonilla N,Durand A,Audemard-Verger A,Guilbert T,Martin B,Lucas B,Auffray C
Published figure using CD73 monoclonal antibody (Product # 12-0731-82) in Flow Cytometry
Thu Dec 14 00:00:00 EST 2017
Three distinct developmental pathways for adaptive and two IFN-γ-producing γδ T subsets in adult thymus.
Nature communications
Buus TB,Ødum N,Geisler C,Lauritsen JPH
Published figure using CD73 monoclonal antibody (Product # 12-0731-82) in Flow Cytometry
Mon Dec 04 00:00:00 EST 2017
Mesenchymal stromal cell-derived extracellular vesicles rescue radiation damage to murine marrow hematopoietic cells.
Leukemia
Wen S,Dooner M,Cheng Y,Papa E,Del Tatto M,Pereira M,Deng Y,Goldberg L,Aliotta J,Chatterjee D,Stewart C,Carpanetto A,Collino F,Bruno S,Camussi G,Quesenberry P
12-0731 was used in Flow cytometry/Cell sorting to evaluate the capacity of MSC-derived extracellular vesicles to mitigate radiation injury to marrow stem cells.
Tue Nov 01 00:00:00 EDT 2016
A New Method to Stabilize C-Kit Expression in Reparative Cardiac Mesenchymal Cells.
Frontiers in cell and developmental biology
Wysoczynski M,Dassanayaka S,Zafir A,Ghafghazi S,Long BW,Noble C,DeMartino AM,Brittian KR,Bolli R,Jones SP
12-0731 was used in Flow cytometry/Cell sorting to investigate a new method for c-kit expressing cell isolation and stabilisation in reparative cardiac mesenchymal cells.
Thu Aug 18 00:00:00 EDT 2016
Developmental Progression and Interrelationship of Central and Effector Regulatory T Cell Subsets.
Journal of immunology (Baltimore, Md. : 1950)
Toomer KH,Yuan X,Yang J,Dee MJ,Yu A,Malek TR
12-0731 was used in Flow cytometry/Cell sorting to elucidate the developmental progression and interrelationship of central and effector regulatory T cell subsets.
Sun May 01 00:00:00 EDT 2016
Accelerated craniofacial bone regeneration through dense collagen gel scaffolds seeded with dental pulp stem cells.
Scientific reports
Chamieh F,Collignon AM,Coyac BR,Lesieur J,Ribes S,Sadoine J,Llorens A,Nicoletti A,Letourneur D,Colombier ML,Nazhat SN,Bouchard P,Chaussain C,Rochefort GY
Published figure using CD73 monoclonal antibody (Product # 12-0731-82) in Flow Cytometry
Fri Dec 09 00:00:00 EST 2016
Limited clonal relatedness between gut IgA plasma cells and memory B cells after oral immunization.
Nature communications
Bemark M,Hazanov H,Strömberg A,Komban R,Holmqvist J,Köster S,Mattsson J,Sikora P,Mehr R,Lycke NY
Published figure using CD73 monoclonal antibody (Product # 12-0731-82) in Flow Cytometry
Tue Sep 06 00:00:00 EDT 2016
Human definitive haemogenic endothelium and arterial vascular endothelium represent distinct lineages.
Nature cell biology
Ditadi A,Sturgeon CM,Tober J,Awong G,Kennedy M,Yzaguirre AD,Azzola L,Ng ES,Stanley EG,French DL,Cheng X,Gadue P,Speck NA,Elefanty AG,Keller G
12-0731 was used in Flow cytometry/Cell sorting to investigate whether haemogenic endothelium represents a subpopulation of arterial vascular endothelium, showing that they represent distinct lineages.
Fri May 01 00:00:00 EDT 2015
Generation of embryonic stem cells from mouse adipose-tissue derived cells via somatic cell nuclear transfer.
Cell cycle (Georgetown, Tex.)
Qin Y,Qin J,Zhou C,Li J,Gao WQ
12-0731 was used in Flow cytometry/Cell sorting to demonstrates that ESCs can be generated from the adipose tissue by somatic cell nuclear transfer (SCNT) and suggests that ADCs can be a new donor-cell type for potential therapeutic cloning.
Tue Jan 26 00:00:00 EST 2016
O-GlcNAcylation Negatively Regulates Cardiomyogenic Fate in Adult Mouse Cardiac Mesenchymal Stromal Cells.
PloS one
Zafir A,Bradley JA,Long BW,Muthusamy S,Li Q,Hill BG,Wysoczynski M,Prabhu SD,Bhatnagar A,Bolli R,Jones SP
12-0731 was used in Flow cytometry/Cell sorting to investigate how O-GlcNAcylation of intracellular proteins is involved in the regulation of mesenchymal stromal cell lineage commitment.
Wed Jun 29 00:00:00 EDT 2016
Cell-autonomous heparanase modulates self-renewal and migration in bone marrow-derived mesenchymal stem cells.
Journal of biomedical science
Cheng CC,Lee YH,Lin SP,Huangfu WC,Liu IH
12-0731 was used in Flow cytometry/Cell sorting to inhibit the enzymatic activity of cell-autonomous HPSE1 in BM-MSCs to clarify the physiological role of HPSE1 in BM-MSCs.
Thu Mar 13 00:00:00 EDT 2014
CD73+ regulatory T cells contribute to adenosine-mediated resolution of acute lung injury.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Ehrentraut H,Clambey ET,McNamee EN,Brodsky KS,Ehrentraut SF,Poth JM,Riegel AK,Westrich JA,Colgan SP,Eltzschig HK
12-0731 was used in Flow cytometry/Cell sorting to determine how CD73-dependent adenosine generation in Tregs promotes ALI resolution.
Sat Jun 01 00:00:00 EDT 2013
A mouse model of adult-onset anaemia due to erythropoietin deficiency.
Nature communications
Yamazaki S,Souma T,Hirano I,Pan X,Minegishi N,Suzuki N,Yamamoto M
12-0731 was used in Immunohistochemistry-immunofluorescence to generate an inherited super-anaemic mice as a mouse model of adult-onset anaemia caused by erythropoietin deficiency.
Mon Dec 30 00:00:00 EST 2013
Multipotent stem cells from trabecular meshwork become phagocytic TM cells.
Investigative ophthalmology & visual science
Du Y,Roh DS,Mann MM,Funderburgh ML,Funderburgh JL,Schuman JS
Wed Mar 21 00:00:00 EDT 2012
Development of murine hepatic NK cells during ontogeny: comparison with spleen NK cells.
Clinical & developmental immunology
Wu X,Chen Y,Wei H,Sun R,Tian Z
12-0731 was used in Flow cytometry/Cell sorting to investigate the phenotype of developing liver NK cells compared to spleen NK cells.
Tue Apr 24 00:00:00 EDT 2012
Adipose-derived stem cells promote lymphangiogenesis in response to VEGF-C stimulation or TGF-β1 inhibition.
Future oncology (London, England)
Yan A,Avraham T,Zampell JC,Haviv YS,Weitman E,Mehrara BJ
12-0731 was used in Flow cytometry/Cell sorting to determine whether ASCs can be used in lymphatic tissue-engineering by altering the balance between lymphangiogenic cytokines.
Thu Dec 01 00:00:00 EST 2011
Adipose-derived stem cells promote lymphangiogenesis in response to VEGF-C stimulation or TGF-β1 inhibition.
Future oncology (London, England)
Yan A,Avraham T,Zampell JC,Haviv YS,Weitman E,Mehrara BJ
12-0731 was used in Flow cytometry/Cell sorting to determine whether ASCs can be used in lymphatic tissue-engineering by altering the balance between lymphangiogenic cytokines.
Thu Dec 01 00:00:00 EST 2011