Product References
Embryonic keratin19+ progenitors generate multiple functionally distinct progeny to maintain epithelial diversity in the adult thymus medulla.
Nature communications
Lucas B,White AJ,Klein F,Veiga-Villauriz C,Handel A,Bacon A,Cosway EJ,James KD,Parnell SM,Ohigashi I,Takahama Y,Jenkinson WE,Hollander GA,Lu WY,Anderson G
Published figure using CD249 (BP-1) monoclonal antibody (Product # 12-5891-82) in Flow Cytometry
Wed Apr 12 00:00:00 EDT 2023
Lymphocyte-Specific Function of the DNA Polymerase Epsilon Subunit Pole3 Revealed by Neomorphic Alleles.
Cell reports
Siamishi I,Iwanami N,Clapes T,Trompouki E,O'Meara CP,Boehm T
12-5891 was used in Flow cytometry/Cell sorting to serve as a paradigm to understand the molecular basis of cell-type-specific non-replicative functions of the ubiquitous POLE complex.
Tue Jun 16 00:00:00 EDT 2020
Epigenetic Protection of Vertebrate Lymphoid Progenitor Cells by Dnmt1.
iScience
Iwanami N,Takeshita K,Lawir DF,Suetake I,Tajima S,Sikora K,Trancoso I,ÓMeara C,Siamishi I,Takahama Y,Furutani-Seiki M,Kondoh H,Yonezawa Y,Schorpp M,Boehm T
12-5891 was used in Flow cytometry/Cell sorting to identify new recessive dnmt1 alleles in medaka and zebrafish and, guided by the structures of mutant proteins, generated a recessive variant of mouse Dnmt1.
Fri Jul 24 00:00:00 EDT 2020
Mutations in topoisomerase IIβ result in a B cell immunodeficiency.
Nature communications
Broderick L,Yost S,Li D,McGeough MD,Booshehri LM,Guaderrama M,Brydges SD,Kucharova K,Patel NC,Harr M,Hakonarson H,Zackai E,Cowell IG,Austin CA,Hügle B,Gebauer C,Zhang J,Xu X,Wang J,Croker BA,Frazer KA,Putnam CD,Hoffman HM
12-5891-82 was used in Flow Cytometry to identify mutations in type II topoisomerase as a driver of defective proliferation and survival of B2 cells, leading to a B cell immunodeficiency.
Tue Aug 13 00:00:00 EDT 2019
Fundamental parameters of the developing thymic epithelium in the mouse.
Scientific reports
Hirakawa M,Nagakubo D,Kanzler B,Avilov S,Krauth B,Happe C,Swann JB,Nusser A,Boehm T
12-5891 was used in Flow cytometry/Cell sorting to study the dynamic of thymic epithelial cells and thymocytes during embryogenesis.
Mon Jul 23 00:00:00 EDT 2018
Lymphatic Contribution to the Cellular Niche in Heterotopic Ossification.
Annals of surgery
Loder S,Agarwal S,Sorkin M,Breuler C,Li J,Peterson J,Gardenier J,Hsieh HH,Wang SC,Mehrara BJ,Levi B
Published figure using CD249 (BP-1) monoclonal antibody (Product # 12-5891-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Distinctive Mesenchymal-Parenchymal Cell Pairings Govern B Cell Differentiation in the Bone Marrow.
Stem cell reports
Yu VW,Lymperi S,Oki T,Jones A,Swiatek P,Vasic R,Ferraro F,Scadden DT
12-5891 was used in Flow cytometry/Cell sorting to study the notion that bone marrow is a composite of specialised niches, providing distinctive functional units to regulate hematopoiesis.
Tue Aug 09 00:00:00 EDT 2016
Mutations in G protein β subunits promote transformation and kinase inhibitor resistance.
Nature medicine
Yoda A,Adelmant G,Tamburini J,Chapuy B,Shindoh N,Yoda Y,Weigert O,Kopp N,Wu SC,Kim SS,Liu H,Tivey T,Christie AL,Elpek KG,Card J,Gritsman K,Gotlib J,Deininger MW,Makishima H,Turley SJ,Javidi-Sharifi N,Maciejewski JP,Jaiswal S,Ebert BL,Rodig SJ,Tyner JW,Marto JA,Weinstock DM,Lane AA
12589182 was used in flow cytometry to investigate the role of GNB1 and GNB2 in cancer cells.
Thu Jan 01 00:00:00 EST 2015
Premature thymic involution is independent of structural plasticity of the thymic stroma.
European journal of immunology
Franckaert D,Schlenner SM,Heirman N,Gill J,Skogberg G,Ekwall O,Put K,Linterman MA,Dooley J,Liston A
12-5891 was used in Flow cytometry/Cell sorting to investigate thymic involution in FVB/N mice, showing that disruption of the epithelial-endothelial relationship and a progressive loss of pro-T cells precedes involution, and loss of pro-T cells is sufficient to drive premature involution, suggesting that pro-T cells are the main driver of involution.
Fri May 01 00:00:00 EDT 2015
Osteoprotegerin-Mediated Homeostasis of Rank+ Thymic Epithelial Cells Does Not Limit Foxp3+ Regulatory T Cell Development.
Journal of immunology (Baltimore, Md. : 1950)
McCarthy NI,Cowan JE,Nakamura K,Bacon A,Baik S,White AJ,Parnell SM,Jenkinson EJ,Jenkinson WE,Anderson G
12-5891 was used in Flow cytometry/Cell sorting to elucidate the conditions controlling medullary thymic epithelial cell (mTEC) homeostasis and the production of mTEC-dependent Foxp3(+) Tregs.
Tue Sep 15 00:00:00 EDT 2015
Triplication of a 21q22 region contributes to B cell transformation through HMGN1 overexpression and loss of histone H3 Lys27 trimethylation.
Nature genetics
Lane AA,Chapuy B,Lin CY,Tivey T,Li H,Townsend EC,van Bodegom D,Day TA,Wu SC,Liu H,Yoda A,Alexe G,Schinzel AC,Sullivan TJ,Malinge S,Taylor JE,Stegmaier K,Jaffe JD,Bustin M,te Kronnie G,Izraeli S,Harris MH,Stevenson KE,Neuberg D,Silverman LB,Sallan SE,Bradner JE,Hahn WC,Crispino JD,Pellman D,Weinstock DM
12-5891 was used in Flow cytometry/Cell sorting to show that germline triplication of only 31 genes orthologous to human chromosome 21q22 confers mouse progenitor B cell self renew in vitro.
Sun Jun 01 00:00:00 EDT 2014
An essential role for medullary thymic epithelial cells during the intrathymic development of invariant NKT cells.
Journal of immunology (Baltimore, Md. : 1950)
White AJ,Jenkinson WE,Cowan JE,Parnell SM,Bacon A,Jones ND,Jenkinson EJ,Anderson G
12-5891 was used in Flow cytometry/Cell sorting to investigate whether thymus microenvironments are required in iNKT cell development, showing that medullary thymic epithelial cells are essential for intrathymic development of iNKT cells.
Sat Mar 15 00:00:00 EDT 2014
Phosphatase Wip1 is essential for the maturation and homeostasis of medullary thymic epithelial cells in mice.
Journal of immunology (Baltimore, Md. : 1950)
Sun L,Li H,Luo H,Zhang L,Hu X,Yang T,Sun C,Chen H,Zhang L,Zhao Y
12-5891 was used in Flow cytometry/Cell sorting to investigate the molecular mechanisms controlling thymic epithelial cell differentiation and activities.
Sun Sep 15 00:00:00 EDT 2013
Developmentally regulated availability of RANKL and CD40 ligand reveals distinct mechanisms of fetal and adult cross-talk in the thymus medulla.
Journal of immunology (Baltimore, Md. : 1950)
Desanti GE,Cowan JE,Baik S,Parnell SM,White AJ,Penninger JM,Lane PJ,Jenkinson EJ,Jenkinson WE,Anderson G
12-5891 was used in Flow cytometry/Cell sorting to study how multiple hemopoietic cell types regulate mTEC development through differential provision of RANKL/CD40L during ontogeny, revealing molecular differences in foetal and adult hemopoietic cross-talk.
Sat Dec 15 00:00:00 EST 2012
Frizzled 9 knock-out mice have abnormal B-cell development.
Blood
Ranheim EA,Kwan HC,Reya T,Wang YK,Weissman IL,Francke U
12-5891 was used in Flow cytometry/Cell sorting to suggest a role for frizzled 9 signalling in lymphoid development.
Tue Mar 15 00:00:00 EST 2005