Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • EAAC1 Antibodies

          Proteintech

          EAAT3 Polyclonal Antibody

          7 References
          View all (10) EAAC1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite EAAT3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          EAAT3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          EAAT3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EAAT3 Antibody (12686-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded mouse brain tissue slide using (Product # 12686-1-AP) (EAAT3 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EAAT3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EAAT3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EAAT3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EAAT3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EAAT3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EAAT3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EAAT3 Antibody in Western Blot (WB)
          EAAT3 Antibody in Western Blot (WB)
          EAAT3 Antibody in Western Blot (WB)
          EAAT3 Antibody in Western Blot (WB)
          EAAT3 Polyclonal Antibody

          Product Details

          12686-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse, Sheep

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          EAAT3 Fusion Protein Ag3379 (430-524 aa encoded by BC033040)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.18 mg/mL

          Amount

          27 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: AEDVTLIIA VDWLLDRFRT MVNVLGDAFG TGIVEKLSKK ELEQMDVSSE VNIVNPFALE STILDNEDSD TKKSYVNGGF AVDKSDTISF TQTSQF (430-524 aa encoded by BC033040)

          Target Information

          L-glutamate is the major excitatory neurotransmitter in mammalian central nervous systems. To maintain glutamate levels below excitoxic levels, excess glutamate at excitatory synaptic clefts is removed by ion-coupled glutamate transporters. Four glutamate transporters have been identified: the sodium-dependent GLAST-1, GLT-1, EAAC-1 and the chloride-dependent EAAT-4. EAAC-1 is expressed in the CNS and also found in kidney and small intestine. Structural features of glutamate transporters are believed to include eight membrane-spanning alpha-helices and a loop-pore structure which is unique among secondary transporters but may resemble loop-pores found in ion channels. A second distinctive structural feature is the presence of a highly amphipathic membrane-spanning helix that provides a hydrophilic path through the membrane. Mice with homozygous deletions of the EAAC-1 transporter gene develop dicarboxylic aminoaciduria and display reduced spontaneous locomotor activity although no neurodegeneration was observed over a period of 12 months.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: excitatory amino acid carrier 1; excitatory amino acid carrier 2; Excitatory amino acid transporter 3; excitatory amino acid transporter-3; Excitatory amino-acid carrier 1; glutamate transporter mEAAC2; Neuronal and epithelial glutamate transporter; Sodium-dependent glutamate/aspartate transporter 3; solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1; Solute carrier family 1 A1 (brain glutamate transporter); Solute carrier family 1 member 1; solute carrier family 1, member 1; unnamed protein product

          View more View less

          Gene Aliases: D130048G10Rik; DCBXA; EAAC1; EAAC2; EAAT3; HEAAC1; MEAAC1; REAAC1; SCZD18; SLC1A1

          View more View less

          UniProt ID: (Human) P43005, (Mouse) P51906, (Rat) P51907

          View more View less

          Entrez Gene ID: (Human) 6505, (Mouse) 20510, (Rat) 25550

          View more View less

          Function(s)
          anion channel activity L-glutamate transmembrane transporter activity high-affinity glutamate transmembrane transporter activity protein binding chloride transmembrane transporter activity L-aspartate transmembrane transporter activity symporter activity glutamate:sodium symporter activity cysteine transmembrane transporter activity identical protein binding metal ion binding D-aspartate transmembrane transporter activity glutamate binding sodium:dicarboxylate symporter activity primary active transporter
          Process(es)
          behavioral fear response response to amphetamine regulation of heart rate glutathione metabolic process glutathione biosynthetic process superoxide metabolic process ion transport neurotransmitter transport amino acid transport cellular zinc ion homeostasis dopamine receptor signaling pathway glutamate receptor signaling pathway chemical synaptic transmission brain development learning or memory memory grooming behavior locomotory behavior response to xenobiotic stimulus positive regulation of heart rate gene expression retina layer formation L-glutamate transport cytokine-mediated signaling pathway neurogenesis adult behavior cellular response to oxidative stress maintenance of blood-brain barrier response to decreased oxygen levels dopamine metabolic process cysteine transport response to morphine negative regulation of neuron apoptotic process establishment of protein localization blood vessel morphogenesis response to axon injury synapse organization neuron apoptotic process L-glutamate import righting reflex retina development in camera-type eye heart contraction long-term synaptic potentiation motor behavior transepithelial transport D-aspartate transmembrane transport L-aspartate transmembrane transport D-aspartate import cellular response to ammonium ion cellular response to mercury ion cellular response to cocaine zinc II ion transmembrane transport response to anesthetic regulation of protein targeting to membrane glutamate homeostasis motor neuron apoptotic process import into cell L-glutamate import across plasma membrane neurotransmitter receptor transport to plasma membrane postsynaptic modulation of chemical synaptic transmission L-aspartate import across plasma membrane transport across blood-brain barrier chloride transmembrane transport cysteine transmembrane transport cellular response to bisphenol A conditioned place preference transport dicarboxylic acid transport sodium ion transmembrane transport protein homooligomerization L-glutamate transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline