Product References
Differentiated mouse kidney tubuloids as a novel in vitro model to study collecting duct physiology.
Frontiers in cell and developmental biology
Olde Hanhof CJA,Dilmen E,Yousef Yengej FA,Latta F,Ammerlaan CME,Schreurs J,Hooijmaijers L,Jansen J,Rookmaaker MB,Orhon I,Verhaar MC,Hoenderop JG
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunohistochemistry (PFA fixed)
Sat Feb 11 00:00:00 EST 2023
Multiplexed evaluation of mouse wound tissue using oligonucleotide barcoding with single-cell RNA sequencing.
STAR protocols
Januszyk M,Griffin M,Mascharak S,Talbott HE,Chen K,Henn D,Spielman AF,Parker JBL,Liang NE,Cotterell A,Guardino N,Foster DS,Wagh D,Coller J,Gurtner GC,Wan DC,Longaker MT
13-3249-82 was used in Flow cytometry/Cell sorting to describe steps to generate skin wounds in mice, followed by tissue harvest and oligonucleotide barcoding.
Fri Mar 17 00:00:00 EDT 2023
Aspirin increases the efficacy of gemcitabine in pancreatic cancer by modulating the PI3K/AKT/mTOR signaling pathway and reversing epithelial‑mesenchymal transition.
Oncology letters
Zhou H,Yun X,Shu Y,Xu K
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Western Blot
Wed Mar 01 00:00:00 EST 2023
Universal Surface Biotinylation: a simple, versatile and cost-effective sample multiplexing method for single-cell RNA-seq analysis.
DNA research : an international journal for rapid publication of reports on genes and genomes
Sugimoto M,Tada Y,Shichino S,Koyamatsu S,Tsumaki N,Abe K
13-3249-82 was used in Flow Cytometry, Immunohistochemistry (Frozen), Western Blot to develop a universal labelling technique, Universal Surface Biotinylation (USB), which does not depend on specific cell surface proteins.
Fri May 27 00:00:00 EDT 2022
Universal Surface Biotinylation: a simple, versatile and cost-effective sample multiplexing method for single-cell RNA-seq analysis.
DNA research : an international journal for rapid publication of reports on genes and genomes
Sugimoto M,Tada Y,Shichino S,Koyamatsu S,Tsumaki N,Abe K
13-3249-82 was used in Flow Cytometry, Immunohistochemistry (Frozen), Western Blot to develop a universal labelling technique, Universal Surface Biotinylation (USB), which does not depend on specific cell surface proteins.
Fri May 27 00:00:00 EDT 2022
Universal Surface Biotinylation: a simple, versatile and cost-effective sample multiplexing method for single-cell RNA-seq analysis.
DNA research : an international journal for rapid publication of reports on genes and genomes
Sugimoto M,Tada Y,Shichino S,Koyamatsu S,Tsumaki N,Abe K
13-3249-82 was used in Flow Cytometry, Immunohistochemistry (Frozen), Western Blot to develop a universal labelling technique, Universal Surface Biotinylation (USB), which does not depend on specific cell surface proteins.
Fri May 27 00:00:00 EDT 2022
Disentangling cadherin-mediated cell-cell interactions in collective cancer cell migration.
Biophysical journal
Zisis T,Brückner DB,Brandstätter T,Siow WX,d'Alessandro J,Vollmar AM,Broedersz CP,Zahler S
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Neutralization
Tue Jan 04 00:00:00 EST 2022
CMTM5 inhibits the development of prostate cancer via the EGFR/PI3K/AKT signaling pathway.
Molecular medicine reports
Li L,Hu Y,Chen D,Zhu J,Bao W,Xu X,Chen H,Chen W,Feng R
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunocytochemistry
Sat Jan 01 00:00:00 EST 2022
Biliverdin/Bilirubin Redox Pair Protects Lens Epithelial Cells against Oxidative Stress in Age-Related Cataract by Regulating NF-κB/iNOS and Nrf2/HO-1 Pathways.
Oxidative medicine and cellular longevity
Huang Y,Li J,Li W,Ai N,Jin H
13-3249-82 was used in Flow Cytometry, Immunohistochemistry (Frozen), Western Blot to reveal that the mRNA expressions of Nrf2, HO-1, and biliverdin reductase A (BVRA) were all decreased in human samples of age-related nuclear cataract and implicate that BV/BR redox pair protects LECs against H2O2-induced apoptosis by regulating NF-κB/iNOS and Nrf2/HO-1 pathways.
Tue Dec 13 00:00:00 EST 2022
Placental superoxide dismutase 3 mediates benefits of maternal exercise on offspring health.
Cell metabolism
Kusuyama J,Alves-Wagner AB,Conlin RH,Makarewicz NS,Albertson BG,Prince NB,Kobayashi S,Kozuka C,Møller M,Bjerre M,Fuglsang J,Miele E,Middelbeek RJW,Xiudong Y,Xia Y,Garneau L,Bhattacharjee J,Aguer C,Patti ME,Hirshman MF,Jessen N,Hatta T,Ovesen PG,Adamo KB,Nozik-Grayck E,Goodyear LJ
13-3249-82 was used in Flow cytometry/Cell sorting to demonstrate that this occurs through a vitamin D receptor-mediated increase in placental superoxide dismutase 3 (SOD3) expression and secretion.
Tue May 04 00:00:00 EDT 2021
Low-Dose Albendazole Inhibits Epithelial-Mesenchymal Transition of Melanoma Cells by Enhancing Phosphorylated GSK-3β/Tyr216 Accumulation.
Journal of oncology
He Z,Lei S,Liang F,Tan L,Zhang W,Xie L,Zheng H,Lu Y
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Western Blot
Fri Dec 31 00:00:00 EST 2021
MicroRNA-384 inhibits nasopharyngeal carcinoma growth and metastasis via binding to Smad5 and suppressing the Wnt/β-catenin axis.
Cytotechnology
Zeng X,Liao H,Wang F
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Western Blot
Thu Apr 01 00:00:00 EDT 2021
Human Adipose-Derived Mesenchymal Stem Cells Ameliorate Elastase-Induced Emphysema in Mice by Mesenchymal-Epithelial Transition.
International journal of chronic obstructive pulmonary disease
Fujioka N,Kitabatake M,Ouji-Sageshima N,Ibaraki T,Kumamoto M,Fujita Y,Hontsu S,Yamauchi M,Yoshikawa M,Muro S,Ito T
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunocytochemistry
Mon Nov 08 00:00:00 EST 2021
Human Adipose-Derived Mesenchymal Stem Cells Ameliorate Elastase-Induced Emphysema in Mice by Mesenchymal-Epithelial Transition.
International journal of chronic obstructive pulmonary disease
Fujioka N,Kitabatake M,Ouji-Sageshima N,Ibaraki T,Kumamoto M,Fujita Y,Hontsu S,Yamauchi M,Yoshikawa M,Muro S,Ito T
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunocytochemistry
Mon Nov 08 00:00:00 EST 2021
EGFR Transgene Stimulates Spontaneous Formation of MCF7 Breast Cancer Cells Spheroids with Partly Loss of HER3 Receptor.
International journal of molecular sciences
Troitskaya O,Novak D,Nushtaeva A,Savinkova M,Varlamov M,Ermakov M,Richter V,Koval O
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunocytochemistry
Mon Nov 29 00:00:00 EST 2021
Crosstalk between E-Cadherin/β-Catenin and NF-κB Signaling Pathways: The Regulation of Host-Pathogen Interaction during Leptospirosis.
International journal of molecular sciences
Hsu SH,Chou LF,Hong CH,Chang MY,Tsai CY,Tian YC,Yang HY,Yang CW
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Neutralization
Sat Dec 04 00:00:00 EST 2021
Opposing effects of Wnt/β-catenin signaling on epithelial and mesenchymal cell fate in the developing cochlea.
Development (Cambridge, England)
Billings SE,Myers NM,Quiruz L,Cheng AG
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunohistochemistry
Tue Jun 01 00:00:00 EDT 2021
Skin-resident immune cells actively coordinate their distribution with epidermal cells during homeostasis.
Nature cell biology
Park S,Matte-Martone C,Gonzalez DG,Lathrop EA,May DP,Pineda CM,Moore JL,Boucher JD,Marsh E,Schmitter-Sánchez A,Cockburn K,Markova O,Bellaïche Y,Greco V
13-3249-82 was used in Flow Cytometry, Immunohistochemistry (Frozen), Western Blot to propose that these cellular mechanisms provide the epidermis with an optimal response to environmental insults.
Sat May 01 00:00:00 EDT 2021
Efficient Messenger RNA Delivery to the Kidney Using Renal Pelvis Injection in Mice.
Pharmaceutics
Oyama N,Kawaguchi M,Itaka K,Kawakami S
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunohistochemistry (PFA fixed)
Fri Oct 29 00:00:00 EDT 2021
Cholesterol Pathway Inhibition Induces TGF-β Signaling to Promote Basal Differentiation in Pancreatic Cancer.
Cancer cell
Gabitova-Cornell L,Surumbayeva A,Peri S,Franco-Barraza J,Restifo D,Weitz N,Ogier C,Goldman AR,Hartman TR,Francescone R,Tan Y,Nicolas E,Shah N,Handorf EA,Cai KQ,O'Reilly AM,Sloma I,Chiaverelli R,Moffitt RA,Khazak V,Fang CY,Golemis EA,Cukierman E,Astsaturov I
13-3249-82 was used in Flow Cytometry to dissect the causal relationship between endogenous cholesterol metabolism and pancreatic ductal adenocarcinoma development and differentiation.
Mon Oct 12 00:00:00 EDT 2020
Epithelial Vegfa Specifies a Distinct Endothelial Population in the Mouse Lung.
Developmental cell
Vila Ellis L,Cain MP,Hutchison V,Flodby P,Crandall ED,Borok Z,Zhou B,Ostrin EJ,Wythe JD,Chen J
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunocytochemistry
Mon Mar 09 00:00:00 EDT 2020
GDNF drives rapid tubule morphogenesis in a novel 3D in vitro model for ADPKD.
Journal of cell science
Dixon EE,Maxim DS,Halperin Kuhns VL,Lane-Harris AC,Outeda P,Ewald AJ,Watnick TJ,Welling PA,Woodward OM
13-3249-80 was used in Immunocytochemistry to isolate the rapid changes in protein localization and gene expression that correlate with altered cell and tubule morphology during cyst initiation.
Thu Jul 16 00:00:00 EDT 2020
GDNF drives rapid tubule morphogenesis in a novel 3D in vitro model for ADPKD.
Journal of cell science
Dixon EE,Maxim DS,Halperin Kuhns VL,Lane-Harris AC,Outeda P,Ewald AJ,Watnick TJ,Welling PA,Woodward OM
13-3249-80 was used in Immunocytochemistry to isolate the rapid changes in protein localization and gene expression that correlate with altered cell and tubule morphology during cyst initiation.
Thu Jul 16 00:00:00 EDT 2020
Leptin Modulates the Metastasis of Canine Inflammatory Mammary Adenocarcinoma Cells Through Downregulation of Lysosomal Protective Protein Cathepsin A (CTSA).
International journal of molecular sciences
Kim JW,Mahiddine FY,Kim GA
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Western Blot
Wed Nov 25 00:00:00 EST 2020
Prrx1 Fibroblasts Represent a Pro-fibrotic Lineage in the Mouse Ventral Dermis.
Cell reports
Leavitt T,Hu MS,Borrelli MR,Januszyk M,Garcia JT,Ransom RC,Mascharak S,desJardins-Park HE,Litzenburger UM,Walmsley GG,Marshall CD,Moore AL,Duoto B,Adem S,Foster DS,Salhotra A,Shen AH,Griffin M,Shen EZ,Barnes LA,Zielins ER,Maan ZN,Wei Y,Chan CKF,Wan DC,Lorenz HP,Chang HY,Gurtner GC,Longaker MT
13-3249-82 was used in Flow Cytometry to identify and characterise a fibroblast subpopulation in the mouse ventral dermis with intrinsic scar-forming potential.
Tue Nov 10 00:00:00 EST 2020
Elucidating the fundamental fibrotic processes driving abdominal adhesion formation.
Nature communications
Foster DS,Marshall CD,Gulati GS,Chinta MS,Nguyen A,Salhotra A,Jones RE,Burcham A,Lerbs T,Cui L,King ME,Titan AL,Ransom RC,Manjunath A,Hu MS,Blackshear CP,Mascharak S,Moore AL,Norton JA,Kin CJ,Shelton AA,Januszyk M,Gurtner GC,Wernig G,Longaker MT
13-3249 was used in Flow cytometry/Cell sorting to present a systematic analysis of mouse and human adhesion tissues.
Thu Aug 13 00:00:00 EDT 2020
Caspase-11 regulates the tumour suppressor function of STAT1 in a murine model of colitis-associated carcinogenesis.
Oncogene
Flood B,Manils J,Nulty C,Flis E,Kenealy S,Barber G,Fay J,Mills KHG,Kay EW,Creagh EM
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunohistochemistry
Mon Apr 01 00:00:00 EDT 2019
Pluripotency reprogramming by competent and incompetent POU factors uncovers temporal dependency for Oct4 and Sox2.
Nature communications
Malik V,Glaser LV,Zimmer D,Velychko S,Weng M,Holzner M,Arend M,Chen Y,Srivastava Y,Veerapandian V,Shah Z,Esteban MA,Wang H,Chen J,Schöler HR,Hutchins AP,Meijsing SH,Pott S,Jauch R
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Flow Cytometry
Fri Aug 02 00:00:00 EDT 2019
Fsp1-Mediated Lineage Tracing Fails to Detect the Majority of Disseminating Cells Undergoing EMT.
Cell reports
Bornes L,van Scheppingen RH,Beerling E,Schelfhorst T,Ellenbroek SIJ,Seinstra D,van Rheenen J
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Flow Cytometry
Tue Nov 26 00:00:00 EST 2019
Defining Reprogramming Checkpoints from Single-Cell Analyses of Induced Pluripotency.
Cell reports
Tran KA,Pietrzak SJ,Zaidan NZ,Siahpirani AF,McCalla SG,Zhou AS,Iyer G,Roy S,Sridharan R
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunocytochemistry
Tue May 07 00:00:00 EDT 2019
Defining Reprogramming Checkpoints from Single-Cell Analyses of Induced Pluripotency.
Cell reports
Tran KA,Pietrzak SJ,Zaidan NZ,Siahpirani AF,McCalla SG,Zhou AS,Iyer G,Roy S,Sridharan R
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunocytochemistry
Tue May 07 00:00:00 EDT 2019
β-Catenin maintains lung epithelial progenitors after lung specification.
Development (Cambridge, England)
Ostrin EJ,Little DR,Gerner-Mauro KN,Sumner EA,Ríos-Corzo R,Ambrosio E,Holt SE,Forcioli-Conti N,Akiyama H,Hanash SM,Kimura S,Huang SXL,Chen J
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunohistochemistry
Fri Mar 09 00:00:00 EST 2018
Differential requirements of androgen receptor in luminal progenitors during prostate regeneration and tumor initiation.
eLife
Chua CW,Epsi NJ,Leung EY,Xuan S,Lei M,Li BI,Bergren SK,Hibshoosh H,Mitrofanova A,Shen MM
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunofluorescence
Mon Jan 15 00:00:00 EST 2018
Differential requirements of androgen receptor in luminal progenitors during prostate regeneration and tumor initiation.
eLife
Chua CW,Epsi NJ,Leung EY,Xuan S,Lei M,Li BI,Bergren SK,Hibshoosh H,Mitrofanova A,Shen MM
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunofluorescence
Mon Jan 15 00:00:00 EST 2018
Identity and dynamics of mammary stem cells during branching morphogenesis.
Nature
Scheele CL,Hannezo E,Muraro MJ,Zomer A,Langedijk NS,van Oudenaarden A,Simons BD,van Rheenen J
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunocytochemistry
Thu Feb 16 00:00:00 EST 2017
Constitutively Active SMAD2/3 Are Broad-Scope Potentiators of Transcription-Factor-Mediated Cellular Reprogramming.
Cell stem cell
Ruetz T,Pfisterer U,Di Stefano B,Ashmore J,Beniazza M,Tian TV,Kaemena DF,Tosti L,Tan W,Manning JR,Chantzoura E,Ottosson DR,Collombet S,Johnsson A,Cohen E,Yusa K,Linnarsson S,Graf T,Parmar M,Kaji K
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Flow Cytometry
Thu Dec 07 00:00:00 EST 2017
Precision-cut Mouse Lung Slices to Visualize Live Pulmonary Dendritic Cells.
Journal of visualized experiments : JoVE
Lyons-Cohen MR,Thomas SY,Cook DN,Nakano H
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunofluorescence
Wed Apr 05 00:00:00 EDT 2017
Quantitative lineage tracing strategies to resolve multipotency in tissue-specific stem cells.
Genes & development
Wuidart A,Ousset M,Rulands S,Simons BD,Van Keymeulen A,Blanpain C
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunofluorescence
Wed Jun 01 00:00:00 EDT 2016
Tumour stroma-derived lipocalin-2 promotes breast cancer metastasis.
The Journal of pathology
Ören B,Urosevic J,Mertens C,Mora J,Guiu M,Gomis RR,Weigert A,Schmid T,Grein S,Brüne B,Jung M
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
Alternative Routes to Induced Pluripotent Stem Cells Revealed by Reprogramming of the Neural Lineage.
Stem cell reports
Jackson SA,Olufs ZP,Tran KA,Zaidan NZ,Sridharan R
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunofluorescence
Tue Mar 08 00:00:00 EST 2016
Alternative Routes to Induced Pluripotent Stem Cells Revealed by Reprogramming of the Neural Lineage.
Stem cell reports
Jackson SA,Olufs ZP,Tran KA,Zaidan NZ,Sridharan R
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunofluorescence
Tue Mar 08 00:00:00 EST 2016
Epithelial-to-mesenchymal transition induces cell cycle arrest and parenchymal damage in renal fibrosis.
Nature medicine
Lovisa S,LeBleu VS,Tampe B,Sugimoto H,Vadnagara K,Carstens JL,Wu CC,Hagos Y,Burckhardt BC,Pentcheva-Hoang T,Nischal H,Allison JP,Zeisberg M,Kalluri R
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Epithelial-to-mesenchymal transition is dispensable for metastasis but induces chemoresistance in pancreatic cancer.
Nature
Zheng X,Carstens JL,Kim J,Scheible M,Kaye J,Sugimoto H,Wu CC,LeBleu VS,Kalluri R
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunohistochemistry
Thu Nov 26 00:00:00 EST 2015
Reprogramming Roadblocks Are System Dependent.
Stem cell reports
Chantzoura E,Skylaki S,Menendez S,Kim SI,Johnsson A,Linnarsson S,Woltjen K,Chambers I,Kaji K
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Flow Cytometry
Tue Sep 08 00:00:00 EDT 2015
Differentiation of human induced pluripotent stem cells to mature functional Purkinje neurons.
Scientific reports
Wang S,Wang B,Pan N,Fu L,Wang C,Song G,An J,Liu Z,Zhu W,Guan Y,Xu ZQ,Chan P,Chen Z,Zhang YA
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunocytochemistry
Wed Mar 18 00:00:00 EDT 2015
Blocking the epithelial-to-mesenchymal transition pathway abrogates resistance to anti-folate chemotherapy in lung cancer.
Cell death & disease
Liang SQ,Marti TM,Dorn P,Froment L,Hall SR,Berezowska S,Kocher G,Schmid RA,Peng RW
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Immunofluorescence
Thu Jul 16 00:00:00 EDT 2015
Single luminal epithelial progenitors can generate prostate organoids in culture.
Nature cell biology
Chua CW,Shibata M,Lei M,Toivanen R,Barlow LJ,Bergren SK,Badani KK,McKiernan JM,Benson MC,Hibshoosh H,Shen MM
Published figure using CD324 (E-Cadherin) monoclonal antibody (Product # 13-3249-82) in Flow Cytometry
Wed Oct 01 00:00:00 EDT 2014
High-resolution analysis with novel cell-surface markers identifies routes to iPS cells.
Nature
O'Malley J,Skylaki S,Iwabuchi KA,Chantzoura E,Ruetz T,Johnsson A,Tomlinson SR,Linnarsson S,Kaji K
13-3249 was used in Flow cytometry/Cell sorting to investigate the routes that fibroblasts take during the generation of induced pluripotent stem cells, showing that there are orderly changes in cell-surface markers.
Thu Jul 04 00:00:00 EDT 2013
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
13-3249 was used in Flow cytometry/Cell sorting to provide data to show the existence of multi-potent mammary progenitor cells that have pregnancy-specific function.
Mon Dec 08 00:00:00 EST 2014
DNA methylation restricts lineage-specific functions of transcription factor Gata4 during embryonic stem cell differentiation.
PLoS genetics
Oda M,Kumaki Y,Shigeta M,Jakt LM,Matsuoka C,Yamagiwa A,Niwa H,Okano M
13-3249 was used in Flow cytometry/Cell sorting to study DNA methylation during embryonic stem cell differentiation.
Sat Jun 01 00:00:00 EDT 2013