Product References
Multimodal CRISPR perturbations of GWAS loci associated with coronary artery disease in vascular endothelial cells.
PLoS genetics
Wünnemann F,Fotsing Tadjo T,Beaudoin M,Lalonde S,Lo KS,Kleinstiver BP,Lettre G
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
Colonization of dermal arterioles by Neisseria meningitidis provides a safe haven from neutrophils.
Nature communications
Manriquez V,Nivoit P,Urbina T,Echenique-Rivera H,Melican K,Fernandez-Gerlinger MP,Flamant P,Schmitt T,Bruneval P,Obino D,Duménil G
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Flow Cytometry
Tue Jul 27 00:00:00 EDT 2021
Differential Characterization of Temozolomide-Resistant Human Glioma Cells.
International journal of molecular sciences
Lai SW,Huang BR,Liu YS,Lin HY,Chen CC,Tsai CF,Lu DY,Lin C
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Flow Cytometry
Tue Jan 02 00:00:00 EST 2018
The critical role of SENP1-mediated GATA2 deSUMOylation in promoting endothelial activation in graft arteriosclerosis.
Nature communications
Qiu C,Wang Y,Zhao H,Qin L,Shi Y,Zhu X,Song L,Zhou X,Chen J,Zhou H,Zhang H,Tellides G,Min W,Yu L
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Immunofluorescence
Thu Jun 01 00:00:00 EDT 2017
The critical role of SENP1-mediated GATA2 deSUMOylation in promoting endothelial activation in graft arteriosclerosis.
Nature communications
Qiu C,Wang Y,Zhao H,Qin L,Shi Y,Zhu X,Song L,Zhou X,Chen J,Zhou H,Zhang H,Tellides G,Min W,Yu L
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Immunofluorescence
Thu Jun 01 00:00:00 EDT 2017
TNF-α enhances vascular cell adhesion molecule-1 expression in human bone marrow mesenchymal stem cells via the NF-κB, ERK and JNK signaling pathways.
Molecular medicine reports
Lu ZY,Chen WC,Li YH,Li L,Zhang H,Pang Y,Xiao ZF,Xiao HW,Xiao Y
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
Endothelial microparticles interact with and support the proliferation of T cells.
Journal of immunology (Baltimore, Md. : 1950)
Wheway J,Latham SL,Combes V,Grau GE
14-1069 was used in Flow cytometry/Cell sorting to explore the effect of endothelial cell microparticles in T cell activation and antigen presentation.
Wed Oct 01 00:00:00 EDT 2014
Cytokine induction of VCAM-1 but not IL13Rα2 on glioma cells: a tale of two antibodies.
PloS one
Mahadev V,Starr R,Wright SL,Martinez C,Jensen MC,Barish ME,Forman SJ,Brown CE
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Flow Cytometry
Mon Dec 22 00:00:00 EST 2014
Cytokine induction of VCAM-1 but not IL13Rα2 on glioma cells: a tale of two antibodies.
PloS one
Mahadev V,Starr R,Wright SL,Martinez C,Jensen MC,Barish ME,Forman SJ,Brown CE
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Flow Cytometry
Mon Dec 22 00:00:00 EST 2014
Cytokine induction of VCAM-1 but not IL13Rα2 on glioma cells: a tale of two antibodies.
PloS one
Mahadev V,Starr R,Wright SL,Martinez C,Jensen MC,Barish ME,Forman SJ,Brown CE
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Flow Cytometry
Mon Dec 22 00:00:00 EST 2014
Cytokine induction of VCAM-1 but not IL13Rα2 on glioma cells: a tale of two antibodies.
PloS one
Mahadev V,Starr R,Wright SL,Martinez C,Jensen MC,Barish ME,Forman SJ,Brown CE
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Flow Cytometry
Mon Dec 22 00:00:00 EST 2014
Effect of nicotine and porphyromonas gingivalis lipopolysaccharide on endothelial cells in vitro.
PloS one
An N,Andrukhov O,Tang Y,Falkensammer F,Bantleon HP,Ouyang X,Rausch-Fan X
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Flow Cytometry
Mon Jun 29 00:00:00 EDT 2015
Comparative evaluation of differentiation potential of menstrual blood- versus bone marrow-derived stem cells into hepatocyte-like cells.
PloS one
Khanjani S,Khanmohammadi M,Zarnani AH,Akhondi MM,Ahani A,Ghaempanah Z,Naderi MM,Eghtesad S,Kazemnejad S
14-1069 was used in Flow cytometry/Cell sorting to investigate hepatic differentiation capacity of menstrual stem cells compared to bone marrow-derived stem cells.
Sat Nov 08 00:00:00 EST 2014
Effects of Aggregatibacter actinomycetemcomitans leukotoxin on endothelial cells.
Microbial pathogenesis
Dietmann A,Millonig A,Combes V,Couraud PO,Kachlany SC,Grau GE
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Immunofluorescence
Wed Feb 12 00:00:00 EST 2014
Effects of Aggregatibacter actinomycetemcomitans leukotoxin on endothelial cells.
Microbial pathogenesis
Dietmann A,Millonig A,Combes V,Couraud PO,Kachlany SC,Grau GE
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 14-1069-82) in Immunofluorescence
Wed Feb 12 00:00:00 EST 2014