Product References
PDGFRβ+ cells play a dual role as hematopoietic precursors and niche cells during mouse ontogeny.
Cell reports
Sá da Bandeira D,Kilpatrick AM,Marques M,Gomez-Salazar M,Ventura T,Gonzalez ZN,Stefancova D,Rossi F,Vermeren M,Vink CS,Beltran M,Henderson NC,Jung B,van der Linden R,van de Werken HJG,van Ijcken WFJ,Betsholtz C,Forbes SJ,Cuervo H,Crisan M
48-0411-82 was used in Flow cytometry/Cell sorting to demonstrate that PDGFRβ+ cells play a dual role in murine hematopoiesis.
Tue Jul 19 00:00:00 EDT 2022
SNAP23 is essential for platelet and mast cell development and required in connective tissue mast cells for anaphylaxis.
The Journal of biological chemistry
Cardenas RA,Gonzalez R,Sanchez E,Ramos MA,Cardenas EI,Rodarte AI,Alcazar-Felix RJ,Isaza A,Burns AR,Heidelberger R,Adachi R
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
The onset of circulation triggers a metabolic switch required for endothelial to hematopoietic transition.
Cell reports
Azzoni E,Frontera V,Anselmi G,Rode C,James C,Deltcheva EM,Demian AS,Brown J,Barone C,Patelli A,Harman JR,Nicholls M,Conway SJ,Morrissey E,Jacobsen SEW,Sparrow DB,Harris AL,Enver T,de Bruijn MFTR
48-0411-82 was used in Immunocytochemistry-immunoflourescence to investigate how and when the onset of circulation affects Hematopoietic stem cells emergence.
Tue Dec 14 00:00:00 EST 2021
All-trans-retinoic acid shifts Th1 towards Th2 cell differentiation by targeting NFAT1 signalling to ameliorate immune-mediated aplastic anaemia.
British journal of haematology
Tang D,Liu S,Sun H,Qin X,Zhou N,Zheng W,Zhang M,Zhou H,Tuersunayi A,Duan C,Chen J
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Immunohistochemistry
Tue Dec 01 00:00:00 EST 2020
Platelets Fuel the Inflammasome Activation of Innate Immune Cells.
Cell reports
Rolfes V,Ribeiro LS,Hawwari I,Böttcher L,Rosero N,Maasewerd S,Santos MLS,Próchnicki T,Silva CMS,Wanderley CWS,Rothe M,Schmidt SV,Stunden HJ,Bertheloot D,Rivas MN,Fontes CJ,Carvalho LH,Cunha FQ,Latz E,Arditi M,Franklin BS
48-0411-82 was used in Flow Cytometry to conclude that platelets provide an additional layer of regulation of inflammasomes and IL-1-driven inflammation.
Tue May 12 00:00:00 EDT 2020
The endothelial protein C receptor plays an essential role in the maintenance of pregnancy.
Science advances
Castillo MM,Yang Q,Sigala AS,McKinney DT,Zhan M,Chen KL,Jarzembowski JA,Sood R
48-0411 was used in Flow cytometry/Cell sorting to investigate the role of thrombotic processes in Placenta-mediated pregnancy complications,.
Sun Nov 01 00:00:00 EDT 2020
Single-nucleotide human disease mutation inactivates a blood-regenerative GATA2 enhancer.
The Journal of clinical investigation
Soukup AA,Zheng Y,Mehta C,Wu J,Liu P,Cao M,Hofmann I,Zhou Y,Zhang J,Johnson KD,Choi K,Keles S,Bresnick EH
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
Iron deficiency disrupts embryonic haematopoiesis but not the endothelial to haematopoietic transition.
Scientific reports
Shvartsman M,Bilican S,Lancrin C
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Tue Apr 23 00:00:00 EDT 2019
The role of platelets in mediating a response to human influenza infection.
Nature communications
Koupenova M,Corkrey HA,Vitseva O,Manni G,Pang CJ,Clancy L,Yao C,Rade J,Levy D,Wang JP,Finberg RW,Kurt-Jones EA,Freedman JE
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Immunocytochemistry
Tue Apr 16 00:00:00 EDT 2019
Adrenergic nerve degeneration in bone marrow drives aging of the hematopoietic stem cell niche.
Nature medicine
Maryanovich M,Zahalka AH,Pierce H,Pinho S,Nakahara F,Asada N,Wei Q,Wang X,Ciero P,Xu J,Leftin A,Frenette PS
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Immunofluorescence
Fri Jun 01 00:00:00 EDT 2018
Large-Scale Clonal Analysis Resolves Aging of the Mouse Hematopoietic Stem Cell Compartment.
Cell stem cell
Yamamoto R,Wilkinson AC,Ooehara J,Lan X,Lai CY,Nakauchi Y,Pritchard JK,Nakauchi H
48-0411 was used in Flow cytometry/Cell sorting to determine the age-related functional changes to the haematopoietic stem cell compartment.
Thu Apr 05 00:00:00 EDT 2018
Transient loss of venous integrity during developmental vascular remodeling leads to red blood cell extravasation and clearance by lymphatic vessels.
Development (Cambridge, England)
Zhang Y,Daubel N,Stritt S,Mäkinen T
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Immunofluorescence
Thu Feb 08 00:00:00 EST 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Tue Mar 20 00:00:00 EDT 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Tue Mar 20 00:00:00 EDT 2018
Differential cytokine contributions of perivascular haematopoietic stem cell niches.
Nature cell biology
Asada N,Kunisaki Y,Pierce H,Wang Z,Fernandez NF,Birbrair A,Ma'ayan A,Frenette PS
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Immunofluorescence
Wed Mar 01 00:00:00 EST 2017
Mediator Kinase Phosphorylation of STAT1 S727 Promotes Growth of Neoplasms With JAK-STAT Activation.
EBioMedicine
Nitulescu II,Meyer SC,Wen QJ,Crispino JD,Lemieux ME,Levine RL,Pelish HE,Shair MD
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
Targeted genome editing restores T cell differentiation in a humanized X-SCID pluripotent stem cell disease model.
Scientific reports
Alzubi J,Pallant C,Mussolino C,Howe SJ,Thrasher AJ,Cathomen T
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Fri Sep 29 00:00:00 EDT 2017
Small molecule p300/catenin antagonist enhances hematopoietic recovery after radiation.
PloS one
Zhao Y,Wu K,Nguyen C,Smbatyan G,Melendez E,Higuchi Y,Chen Y,Kahn M
48-0411 was used in Flow cytometry/Cell sorting to report that the small molecule YH250 stimulates hematopoiesis in lethally or sublethally irradiated mice.
Tue Sep 26 00:00:00 EDT 2017
Hematopoietic Stem Cells Count and Remember Self-Renewal Divisions.
Cell
Bernitz JM,Kim HS,MacArthur B,Sieburg H,Moore K
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Thu Nov 17 00:00:00 EST 2016
The Poly(C) Binding Protein Pcbp2 and Its Retrotransposed Derivative Pcbp1 Are Independently Essential to Mouse Development.
Molecular and cellular biology
Ghanem LR,Kromer A,Silverman IM,Chatterji P,Traxler E,Penzo-Mendez A,Weiss MJ,Stanger BZ,Liebhaber SA
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Fri Jan 15 00:00:00 EST 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Activation of the TGFβ pathway impairs endothelial to haematopoietic transition.
Scientific reports
Vargel Ö,Zhang Y,Kosim K,Ganter K,Foehr S,Mardenborough Y,Shvartsman M,Enright AJ,Krijgsveld J,Lancrin C
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Fri Feb 19 00:00:00 EST 2016
Small Molecule Inhibitor of CBFβ-RUNX Binding for RUNX Transcription Factor Driven Cancers.
EBioMedicine
Illendula A,Gilmour J,Grembecka J,Tirumala VSS,Boulton A,Kuntimaddi A,Schmidt C,Wang L,Pulikkan JA,Zong H,Parlak M,Kuscu C,Pickin A,Zhou Y,Gao Y,Mishra L,Adli M,Castilla LH,Rajewski RA,Janes KA,Guzman ML,Bonifer C,Bushweller JH
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
Novel function of FAXDC2 in megakaryopoiesis.
Blood cancer journal
Jin Q,Ren Y,Wang M,Suraneni PK,Li D,Crispino JD,Fan J,Huang Z
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Fri Sep 30 00:00:00 EDT 2016
EphrinB2 regulates the emergence of a hemogenic endothelium from the aorta.
Scientific reports
Chen II,Caprioli A,Ohnuki H,Kwak H,Porcher C,Tosato G
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Immunocytochemistry
Thu Jun 02 00:00:00 EDT 2016
EphrinB2 regulates the emergence of a hemogenic endothelium from the aorta.
Scientific reports
Chen II,Caprioli A,Ohnuki H,Kwak H,Porcher C,Tosato G
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Immunocytochemistry
Thu Jun 02 00:00:00 EDT 2016
EphrinB2 regulates the emergence of a hemogenic endothelium from the aorta.
Scientific reports
Chen II,Caprioli A,Ohnuki H,Kwak H,Porcher C,Tosato G
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Immunocytochemistry
Thu Jun 02 00:00:00 EDT 2016
Reprogramming mouse fibroblasts into engraftable myeloerythroid and lymphoid progenitors.
Nature communications
Cheng H,Ang HY,A El Farran C,Li P,Fang HT,Liu TM,Kong SL,Chin ML,Ling WY,Lim EK,Li H,Huber T,Loh KM,Loh YH,Lim B
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Mon Nov 21 00:00:00 EST 2016
H2S protects against fatal myelosuppression by promoting the generation of megakaryocytes/platelets.
Journal of hematology & oncology
Liu HD,Zhang AJ,Xu JJ,Chen Y,Zhu YC
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Wed Feb 24 00:00:00 EST 2016
H2S protects against fatal myelosuppression by promoting the generation of megakaryocytes/platelets.
Journal of hematology & oncology
Liu HD,Zhang AJ,Xu JJ,Chen Y,Zhu YC
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Wed Feb 24 00:00:00 EST 2016
Pak2 restrains endomitosis during megakaryopoiesis and alters cytoskeleton organization.
Blood
Kosoff RE,Aslan JE,Kostyak JC,Dulaimi E,Chow HY,Prudnikova TY,Radu M,Kunapuli SP,McCarty OJ,Chernoff J
48-0411 was used in Flow cytometry/Cell sorting to identify a novel role for Pak2 as an important regulator of megakaryopoiesis, polyploidization and cytoskeletal mechanics in developing megakaryocytes.
Thu May 07 00:00:00 EDT 2015
FOXF1 inhibits hematopoietic lineage commitment during early mesoderm specification.
Development (Cambridge, England)
Fleury M,Eliades A,Carlsson P,Lacaud G,Kouskoff V
48-0411 was used in Flow cytometry/Cell sorting to investigate the lineage specification of early mesodermal precursors expressing or not the Forkhead transcription factor FOXF1.
Thu Oct 01 00:00:00 EDT 2015
Specific Inflammatory Stimuli Lead to Distinct Platelet Responses in Mice and Humans.
PloS one
Beaulieu LM,Clancy L,Tanriverdi K,Benjamin EJ,Kramer CD,Weinberg EO,He X,Mekasha S,Mick E,Ingalls RR,Genco CA,Freedman JE
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Fri Mar 25 00:00:00 EDT 2016
A Quantitative Proteomic Analysis of Hemogenic Endothelium Reveals Differential Regulation of Hematopoiesis by SOX17.
Stem cell reports
Clarke RL,Robitaille AM,Moon RT,Keller G
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Tue Aug 11 00:00:00 EDT 2015
Lysophosphatidic acid acts as a nutrient-derived developmental cue to regulate early hematopoiesis.
The EMBO journal
Li H,Yue R,Wei B,Gao G,Du J,Pei G
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Tue Jun 17 00:00:00 EDT 2014
Platelet glycoprotein Ib-IX as a regulator of systemic inflammation.
Arteriosclerosis, thrombosis, and vascular biology
Corken A,Russell S,Dent J,Post SR,Ware J
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Thu May 01 00:00:00 EDT 2014
Experimental cerebral malaria pathogenesis--hemodynamics at the blood brain barrier.
PLoS pathogens
Nacer A,Movila A,Sohet F,Girgis NM,Gundra UM,Loke P,Daneman R,Frevert U
48-0411 was used in Immunofluorescence to show that significantly more CD8(+) T cells, neutrophils, and macrophages are recruited to postcapillary venules during ECM compared to hyperparasitemia.
Mon Dec 01 00:00:00 EST 2014
Runx1 is required for progression of CD41+ embryonic precursors into HSCs but not prior to this.
Development (Cambridge, England)
Liakhovitskaia A,Rybtsov S,Smith T,Batsivari A,Rybtsova N,Rode C,de Bruijn M,Buchholz F,Gordon-Keylock S,Zhao S,Medvinsky A
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Runx1 is required for progression of CD41+ embryonic precursors into HSCs but not prior to this.
Development (Cambridge, England)
Liakhovitskaia A,Rybtsov S,Smith T,Batsivari A,Rybtsova N,Rode C,de Bruijn M,Buchholz F,Gordon-Keylock S,Zhao S,Medvinsky A
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Runx1 is required for progression of CD41+ embryonic precursors into HSCs but not prior to this.
Development (Cambridge, England)
Liakhovitskaia A,Rybtsov S,Smith T,Batsivari A,Rybtsova N,Rode C,de Bruijn M,Buchholz F,Gordon-Keylock S,Zhao S,Medvinsky A
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Single-cell analyses of regulatory network perturbations using enhancer-targeting TALEs suggest novel roles for PU.1 during haematopoietic specification.
Development (Cambridge, England)
Wilkinson AC,Kawata VK,Schütte J,Gao X,Antoniou S,Baumann C,Woodhouse S,Hannah R,Tanaka Y,Swiers G,Moignard V,Fisher J,Hidetoshi S,Tijssen MR,de Bruijn MF,Liu P,Göttgens B
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Wed Oct 01 00:00:00 EDT 2014
A crucial role for the ubiquitously expressed transcription factor Sp1 at early stages of hematopoietic specification.
Development (Cambridge, England)
Gilmour J,Assi SA,Jaegle U,Kulu D,van de Werken H,Clarke D,Westhead DR,Philipsen S,Bonifer C
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Sun Jun 01 00:00:00 EDT 2014
CD41 is a reliable identification and activation marker for murine basophils in the steady state and during helminth and malarial infections.
European journal of immunology
Bakocevic N,Claser C,Yoshikawa S,Jones LA,Chew S,Goh CC,Malleret B,Larbi A,Ginhoux F,de Lafaille MC,Karasuyama H,Renia L,Ng LG
48-0411 was used in Flow cytometry/Cell sorting to study the CD41 expression within basophils from the blood during both steady state and malarial inections within mice.
Sun Jun 01 00:00:00 EDT 2014
Expression of podocalyxin separates the hematopoietic and vascular potentials of mouse embryonic stem cell-derived mesoderm.
Stem cells (Dayton, Ohio)
Zhang H,Nieves JL,Fraser ST,Isern J,Douvaras P,Papatsenko D,D'Souza SL,Lemischka IR,Dyer MA,Baron MH
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2014
PSTPIP2 dysregulation contributes to aberrant terminal differentiation in GATA-1-deficient megakaryocytes by activating LYN.
Cell death & disease
Liu L,Wen Q,Gong R,Gilles L,Stankiewicz MJ,Li W,Guo M,Li L,Sun X,Li W,Crispino JD,Huang Z
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Thu Jan 09 00:00:00 EST 2014
A directional switch of integrin signalling and a new anti-thrombotic strategy.
Nature
Shen B,Zhao X,O'Brien KA,Stojanovic-Terpo A,Delaney MK,Kim K,Cho J,Lam SC,Du X
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Thu Nov 07 00:00:00 EST 2013
A directional switch of integrin signalling and a new anti-thrombotic strategy.
Nature
Shen B,Zhao X,O'Brien KA,Stojanovic-Terpo A,Delaney MK,Kim K,Cho J,Lam SC,Du X
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Thu Nov 07 00:00:00 EST 2013
Cathepsin G-dependent modulation of platelet thrombus formation in vivo by blood neutrophils.
PloS one
Faraday N,Schunke K,Saleem S,Fu J,Wang B,Zhang J,Morrell C,Dore S
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Mon Apr 14 00:00:00 EDT 2014
Cis-element mutated in GATA2-dependent immunodeficiency governs hematopoiesis and vascular integrity.
The Journal of clinical investigation
Johnson KD,Hsu AP,Ryu MJ,Wang J,Gao X,Boyer ME,Liu Y,Lee Y,Calvo KR,Keles S,Zhang J,Holland SM,Bresnick EH
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2012
Neuroimmunological blood brain barrier opening in experimental cerebral malaria.
PLoS pathogens
Nacer A,Movila A,Baer K,Mikolajczak SA,Kappe SH,Frevert U
48-0411 was used in in vivo experiments to better understand disease progression in experimental cerebral malaria.
Thu Apr 04 00:00:00 EDT 2013
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
Signaling profiling at the single-cell level identifies a distinct signaling signature in murine hematopoietic stem cells.
Stem cells (Dayton, Ohio)
Du J,Wang J,Kong G,Jiang J,Zhang J,Liu Y,Tong W,Zhang J
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2012
ER71 directs mesodermal fate decisions during embryogenesis.
Development (Cambridge, England)
Rasmussen TL,Kweon J,Diekmann MA,Belema-Bedada F,Song Q,Bowlin K,Shi X,Ferdous A,Li T,Kyba M,Metzger JM,Koyano-Nakagawa N,Garry DJ
Published figure using CD41a monoclonal antibody (Product # 48-0411-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2011
Acute systemic reaction and lung alterations induced by an antiplatelet integrin gpIIb/IIIa antibody in mice.
Blood
Nieswandt B,Echtenacher B,Wachs FP,Schröder J,Gessner JE,Schmidt RE,Grau GE,Männel DN
Thu Jul 15 00:00:00 EDT 1999
Acute systemic reaction and lung alterations induced by an antiplatelet integrin gpIIb/IIIa antibody in mice.
Blood
Nieswandt B,Echtenacher B,Wachs FP,Schröder J,Gessner JE,Schmidt RE,Grau GE,Männel DN
Thu Jul 15 00:00:00 EDT 1999
Acute systemic reaction and lung alterations induced by an antiplatelet integrin gpIIb/IIIa antibody in mice.
Blood
Nieswandt B,Echtenacher B,Wachs FP,Schröder J,Gessner JE,Schmidt RE,Grau GE,Männel DN
Thu Jul 15 00:00:00 EDT 1999
Acute systemic reaction and lung alterations induced by an antiplatelet integrin gpIIb/IIIa antibody in mice.
Blood
Nieswandt B,Echtenacher B,Wachs FP,Schröder J,Gessner JE,Schmidt RE,Grau GE,Männel DN
Thu Jul 15 00:00:00 EDT 1999