Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: This GR-P monoclonal antibody reacts with human CD42a, which is also known as glycoprotein IX (GPIX). CD42a is a 22-kDa transmembrane protein that is constitutively expressed on platelets and megakaryocytes. CD42a associates with CD42b, CD42c, and CD42d to form the GPIb-V-IX complex, which binds the von Willebrand factor, an endothelial adhesion protein, to mediate platelet adhesion and activation. This binding results in platelet aggregation at sites of vascular injury. This receptor complex has also been demonstrated to bind thrombin, factors XI and XII, P-selectin, and Mac-1. Finally, a point mutation within the leucine-rich motif of CD42a has been linked to Bernard-Soulier syndrome.
Applications Reported: Anti-Human CD42a eFluor® 450 has been reported for use in flow cytometric analysis.
Applications Tested: The GR-P antibody has been pre-titrated and tested by flow cytometric analysis of normal human platelets. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.
Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Single-chain membrane glycoprotein that forms a non-covalent complex with GPIb. (MW 23 kDa) Reactivity with resting and activated platelets, weakly on monocytes, megakaryocytes and attachment site for the platelet plasma membrane to the submembrane cytoskeleton. GPIb/IX complex, functions as the receptor for ristocetin-induced binding of von Willebrand factor and as the von Willebrand factor-depend adhesion receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD42a; Glycoprotein 9; glycoprotein IX (platelet); GP-IX; Platelet glycoprotein IX; Platelet GPIX
Gene Aliases: CD42a; GP9; GPIX
UniProt ID: (Human) P14770
Entrez Gene ID: (Human) 2815
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support