Product References
Dynamics of CXCR4 positive circulating tumor cells in prostate cancer patients during radiotherapy.
International journal of cancer
Klusa D,Lohaus F,Franken A,Baumbach M,Cojoc M,Dowling P,Linge A,Offermann A,Löck S,Hušman D,Rivandi M,Polzer B,Freytag V,Lange T,Neubauer H,Kücken M,Perner S,Hölscher T,Dubrovska A,Krause M,Kurth I,Baumann M,Peitzsch C
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Thu Jun 15 00:00:00 EDT 2023
Parathyroid Hormone-Related Protein Inhibition Blocks Triple-Negative Breast Cancer Expansion in Bone Through Epithelial to Mesenchymal Transition Reversal.
JBMR plus
Li J,Camirand A,Zakikhani M,Sellin K,Guo Y,Luan X,Mihalcioiu C,Kremer R
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Immunocytochemistry
Wed Jun 01 00:00:00 EDT 2022
The sodium/glucose cotransporters as potential therapeutic targets for CF lung diseases revealed by human lung organoid swelling assay.
Molecular therapy. Methods & clinical development
Hirai H,Liang X,Sun Y,Zhang Y,Zhang J,Chen YE,Mou H,Zhao Y,Xu J
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Thu Mar 10 00:00:00 EST 2022
Tumor suppressor DEAR1 regulates mammary epithelial cell fate and predicts early onset and metastasis in triple negative breast cancer.
Scientific reports
Le UQ,Chen N,Balasenthil S,Lurie E,Yang F,Liu S,Rubin L,Solis Soto LM,Raso MG,Batra H,Sahin AA,Wistuba II,Killary AM
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Mon Nov 14 00:00:00 EST 2022
Tumor suppressor DEAR1 regulates mammary epithelial cell fate and predicts early onset and metastasis in triple negative breast cancer.
Scientific reports
Le UQ,Chen N,Balasenthil S,Lurie E,Yang F,Liu S,Rubin L,Solis Soto LM,Raso MG,Batra H,Sahin AA,Wistuba II,Killary AM
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Mon Nov 14 00:00:00 EST 2022
The Effect of Infant Gastric Digestion on Human Maternal Milk Cells.
Molecular nutrition & food research
Doerfler R,Melamed JR,Whitehead KA
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Sat Oct 01 00:00:00 EDT 2022
EMP3 negatively modulates breast cancer cell DNA replication, DNA damage repair, and stem-like properties.
Cell death & disease
Zhou K,Sun Y,Dong D,Zhao C,Wang W
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Sun Sep 12 00:00:00 EDT 2021
Helicobacter pylori promote inflammation and host defense through the cagA-dependent activation of mTORC1.
Journal of cellular physiology
Feng GJ,Chen Y,Li K
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Immunocytochemistry
Tue Dec 01 00:00:00 EST 2020
FZD5 contributes to TNBC proliferation, DNA damage repair and stemness.
Cell death & disease
Sun Y,Wang Z,Na L,Dong D,Wang W,Zhao C
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Sat Dec 12 00:00:00 EST 2020
Chemotherapeutic Stress Influences Epithelial-Mesenchymal Transition and Stemness in Cancer Stem Cells of Triple-Negative Breast Cancer.
International journal of molecular sciences
Li X,Strietz J,Bleilevens A,Stickeler E,Maurer J
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Wed Jan 08 00:00:00 EST 2020
Profiling human breast epithelial cells using single cell RNA sequencing identifies cell diversity.
Nature communications
Nguyen QH,Pervolarakis N,Blake K,Ma D,Davis RT,James N,Phung AT,Willey E,Kumar R,Jabart E,Driver I,Rock J,Goga A,Khan SA,Lawson DA,Werb Z,Kessenbrock K
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Wed May 23 00:00:00 EDT 2018
TGF-β1 secreted by Tregs in lymph nodes promotes breast cancer malignancy via up-regulation of IL-17RB.
EMBO molecular medicine
Huang SC,Wei PC,Hwang-Verslues WW,Kuo WH,Jeng YM,Hu CM,Shew JY,Huang CS,Chang KJ,Lee EY,Lee WH
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
TGF-β1 secreted by Tregs in lymph nodes promotes breast cancer malignancy via up-regulation of IL-17RB.
EMBO molecular medicine
Huang SC,Wei PC,Hwang-Verslues WW,Kuo WH,Jeng YM,Hu CM,Shew JY,Huang CS,Chang KJ,Lee EY,Lee WH
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
KDM4 Inhibition Targets Breast Cancer Stem-like Cells.
Cancer research
Metzger E,Stepputtis SS,Strietz J,Preca BT,Urban S,Willmann D,Allen A,Zenk F,Iovino N,Bronsert P,Proske A,Follo M,Boerries M,Stickeler E,Xu J,Wallace MB,Stafford JA,Kanouni T,Maurer J,Schüle R
50-9326 was used in Flow cytometry/Cell sorting to validate a BCSC culture system for drug screening and proof of concept for KDM4 inhibition as a triple-negative breast cancer treatment.
Wed Nov 01 00:00:00 EDT 2017
Evidence of In Vitro Preservation of Human Nephrogenesis at the Single-Cell Level.
Stem cell reports
Pode-Shakked N,Gershon R,Tam G,Omer D,Gnatek Y,Kanter I,Oriel S,Katz G,Harari-Steinberg O,Kalisky T,Dekel B
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Tue Jul 11 00:00:00 EDT 2017
Evidence of In Vitro Preservation of Human Nephrogenesis at the Single-Cell Level.
Stem cell reports
Pode-Shakked N,Gershon R,Tam G,Omer D,Gnatek Y,Kanter I,Oriel S,Katz G,Harari-Steinberg O,Kalisky T,Dekel B
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Tue Jul 11 00:00:00 EDT 2017
Epithelia Use Butyrophilin-like Molecules to Shape Organ-Specific γδ T Cell Compartments.
Cell
Di Marco Barros R,Roberts NA,Dart RJ,Vantourout P,Jandke A,Nussbaumer O,Deban L,Cipolat S,Hart R,Iannitto ML,Laing A,Spencer-Dene B,East P,Gibbons D,Irving PM,Pereira P,Steinhoff U,Hayday A
50-9326 was used in Flow cytometry/Cell sorting to investigate the mechanism that shape organ-specific γδ T cell compartments, showing that Btnl1 expressed by murine enterocytes shapes the local γδ compartment, as does BTNL3 and BTNL8 expressed by human gut epithelial cells, suggesting a conserved mechanism shaped local T cell compartments.
Thu Sep 22 00:00:00 EDT 2016
Clostridium perfringens enterotoxin C-terminal domain labeled to fluorescent dyes for in vivo visualization of micrometastatic chemotherapy-resistant ovarian cancer.
International journal of cancer
Cocco E,Shapiro EM,Gasparrini S,Lopez S,Schwab CL,Bellone S,Bortolomai I,Sumi NJ,Bonazzoli E,Nicoletti R,Deng Y,Saltzman WM,Zeiss CJ,Centritto F,Black JD,Silasi DA,Ratner E,Azodi M,Rutherford TJ,Schwartz PE,Pecorelli S,Santin AD
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Immunofluorescence
Tue Dec 01 00:00:00 EST 2015
The isolation and characterization of CTC subsets related to breast cancer dormancy.
Scientific reports
Vishnoi M,Peddibhotla S,Yin W,T Scamardo A,George GC,Hong DS,Marchetti D
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Thu Dec 03 00:00:00 EST 2015
NDY1/KDM2B functions as a master regulator of polycomb complexes and controls self-renewal of breast cancer stem cells.
Cancer research
Kottakis F,Foltopoulou P,Sanidas I,Keller P,Wronski A,Dake BT,Ezell SA,Shen Z,Naber SP,Hinds PW,McNiel E,Kuperwasser C,Tsichlis PN
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Tue Jul 15 00:00:00 EDT 2014
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
Estrogen and progesterone together expand murine endometrial epithelial progenitor cells.
Stem cells (Dayton, Ohio)
Janzen DM,Cheng D,Schafenacker AM,Paik DY,Goldstein AS,Witte ON,Jaroszewicz A,Pellegrini M,Memarzadeh S
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Immunohistochemistry
Mon Apr 01 00:00:00 EDT 2013
Stem-like epithelial cells are concentrated in the distal end of the fallopian tube: a site for injury and serous cancer initiation.
Stem cells (Dayton, Ohio)
Paik DY,Janzen DM,Schafenacker AM,Velasco VS,Shung MS,Cheng D,Huang J,Witte ON,Memarzadeh S
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Thu Nov 01 00:00:00 EDT 2012
Stem-like epithelial cells are concentrated in the distal end of the fallopian tube: a site for injury and serous cancer initiation.
Stem cells (Dayton, Ohio)
Paik DY,Janzen DM,Schafenacker AM,Velasco VS,Shung MS,Cheng D,Huang J,Witte ON,Memarzadeh S
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Thu Nov 01 00:00:00 EDT 2012
Slow-cycling therapy-resistant cancer cells.
Stem cells and development
Moore N,Houghton J,Lyle S
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 50-9326-42) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2012