Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Description: This 1H7 monoclonal antibody reacts with human insulin-like growth factor-1 receptor (IGF-1R), also known as CD221. This receptor is a cell surface-expressed glycoprotein composed of two extracellular alpha subunits and two transmembrane beta subunits that possess tyrosine kinase activity. Expressed on nearly all cell types, this receptor binds IGF-I and IGF-II, as well as insulin. Ligand binding leads to activation of the PI3K/Akt and MAPK pathways, which mediate cell proliferation and survival. Many tumors and transformed cells display altered IGF-1R expression.
This monoclonal antibody has been reported to block binding of IGF-I and IGF-II to the IGF-1R.
Applications Reported: This 1H7 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This 1H7 antibody has been pre-titrated and tested by flow cytometric analysis of the HeLa cell line. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.
Filtration: 0.2 µm post-manufacturing filtered.
IGF1R, also known as Insulin-like Growth Factor 1 (IGF-1) Receptor, is a receptor tyrosine kinase activated by IGF-1 and IGF-2. IGFR has been implicated in a variety of cancers. The protein is a tetramer consisting of two alpha, and two beta subunits. The alpha-subunits binds to insulin growth factor 1 with higher affinity than to insulin growth factor 2. Binding results in a conformational change, followed by autophosphorylation of IGF-1R at positions 1134, 1135, 1136. The activated IGF-1R in turn, causes phosphorylation of substrate proteins, followed by sequential activation of RAS, RAF, and mitogen-activated protein kinase isoforms ERK, p38, and JNK, leading to the transcription of genes that drive proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support