Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Abl2 Antibodies

          Abnova

          ABL2 Polyclonal Antibody, MaxPab™

          View all (26) Abl2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ABL2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ABL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ABL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ABL2 Antibody (H00000027-B01P) in WB

          Western Blot analysis of ABL2 expression in transfected 293T cell line by ABL2 MaxPab polyclonal antibody. Lane 1: ABL2 transfected lysate(126.70 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ABL2 Antibody in Western Blot (WB)
          ABL2 Polyclonal Antibody, MaxPab™

          Product Details

          H00000027-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ABL2 (AAH65912.1, 1 a.a. approximately 1167 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MVLGTVLLPP NSYGRDQDTS LCCLCTEASE SALPDLTDHF ASCVEDGFEG DKTGGSSPEA LHRPYGCDVE PQALNEAIRW SSKENLLGAT ESDPNLFVAL YDFVASGDNT LSITKGEKLR VLGYNQNGEW SEVRSKNGQG WVPSNYITPV NSLEKHSWYH GPVSRSAAEY LLSSLINGSF LVRESESSPG QLSISLRYEG RVYHYRINTT ADGKVYVTAE SRFSTLAELV HHHSTVADGL VTTLHYPAPK CNKPTVYGVS PIHDKWEMER TDITMKHKLG GGQYGEVYVG VWKKYSLTVA VKTLKEDTME VEEFLKEAAV MKEIKHPNLV QLLGVCTLEP PFYIVTEYMP YGNLLDYLRE CNREEVTAVV LLYMATQISS AMEYLEKKNF IHRDLAARNC LVGENHVVKV ADFGLSRLMT GDTYTAHAGA KFPIKWTAPE SLAYNTFSIK SDVWAFGVLL WEIATYGMSP YPGIDLSQVY DLLEKGYRME QPEGCPPKVY ELMRACWKWS PADRPSFAET HQAFETMFHD SSISEEVAEE LGRAASSSSV VPYLPRLPIL PSKTRTLKKQ VENKENIEGA QDATENSASS LAPGFIRGAQ ASSGSPALPR KQRDKSPSSL LEDAKETCFT RDRKGGFFSS FMKKRNAPTP PKRSSSFREM ENQPHKKYEL TGNFSSVASL QHADGFSFTP AQQEANLVPP KCYGGSFAQR NLCNDDGGGG GGSGTAGGGW SGITGFFTPR LIKKTLGLRA GKPTASDDTS KPFPRSNSTS SMSSGLPEQD RMAMTLPRNC QRSKLQLERT VSTSSQPEEN VDRANDMLPK KSEESAAPSR ERPKAKLLPR GATALPLRTP SGDLAITEKD PPGVGVAGVA AAPKGKEKNG GARLGMAGVP EDGEQPGWPS PAKAAPVLPT THNHKVPVLI SPTLKHTPAD VQLIGTDSQG NKFKLLSEHQ VTSSGDKDRP RRVKPKCAPP PPPVMRLLQH PSICSDPTEE PTALTAGQST SETQEGGKKA ALGAVPISGK AGRPVMPPPQ VPLPTSSISP AKMANGTAGT KVALRKTKQA AEKISADKIS KEALLECADL LSSALTEPVP NSQLVDTGHQ LLDYCSGYVD CIPQTRNKFA FREAVSKLEL SLQELQVSSA AAGVPGTNPV LNNLLSCVQE ISDVVQR

          Target Information

          ABL2 (Arg) is an Abl-related non-receptor protein tyrosine kinase that links cell surface receptors to cell proliferation and/or cytoskeletal reorganization. Abl2 encodes a member of the Abelson family of nonreceptor tyrosine protein kinase. The protein is highly similar to the ABL1 protein, including the tyrosine kinase, SH2 and SH3 domains, and has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. Abl2 is expressed in both normal and tumor cells, and is involved in translocation with the ETV6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Abelson murine leukemia viral oncogene homolog 2; Abelson tyrosine-protein kinase 2; Abelson-related gene protein; arg tyrosine kinase; c-abl oncogene 2, non-receptor tyrosine kinase; FLJ22224; FLJ31718; FLJ41441; Tyrosine-protein kinase ABL2; Tyrosine-protein kinase ARG; v-abl Abelson murine leukemia viral oncogene homolog 2

          View more View less

          Gene Aliases: ABL2; ABLL; ARG

          View more View less

          UniProt ID: (Human) P42684

          View more View less

          Entrez Gene ID: (Human) 27

          View more View less

          Function(s)
          nucleotide binding magnesium ion binding phosphotyrosine binding actin monomer binding protein kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity protein binding ATP binding enzyme activator activity kinase activity transferase activity enzyme binding manganese ion binding metal ion binding actin filament binding
          Process(es)
          cell adhesion signal transduction epidermal growth factor receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration regulation of autophagy positive regulation of neuron projection development peptidyl-tyrosine phosphorylation regulation of endocytosis regulation of cell adhesion regulation of actin cytoskeleton organization cellular response to oxidative stress negative regulation of Rho protein signal transduction exploration behavior protein modification process cellular response to retinoic acid positive regulation of establishment of T cell polarity regulation of cell motility positive regulation of T cell migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline