Product References
Regulation of IgE production and airway reactivity by CD4⁻CD8⁻ regulatory T cells.
Immunobiology
Raker V,Stein J,Montermann E,Maxeiner J,Taube C,Reske-Kunz AB,Sudowe S
HM3500 was used in flow cytometry to characterize high zone tolerance in mouse models of type I allergy and of allergic airway inflammation.
Wed Apr 01 00:00:00 EDT 2015
Pretransplant infusion of mesenchymal stem cells prolongs the survival of a semiallogeneic heart transplant through the generation of regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Casiraghi F,Azzollini N,Cassis P,Imberti B,Morigi M,Cugini D,Cavinato RA,Todeschini M,Solini S,Sonzogni A,Perico N,Remuzzi G,Noris M
HM3500 was used in blocking or activating experiment to test if mesenchymal stem cells have immunomodulatory properties in solid organ allotransplantation
Mon Sep 15 00:00:00 EDT 2008
Cyclin-dependent kinase inhibition limits glomerulonephritis and extends lifespan of mice with systemic lupus.
Arthritis and rheumatism
Zoja C,Casiraghi F,Conti S,Corna D,Rottoli D,Cavinato RA,Remuzzi G,Benigni A
HM3500 was used in blocking or activating experiment to study the effects of CDK inhibition on glomerulonephritis and lifespan of mice with systemic lupus
Tue May 01 00:00:00 EDT 2007
IL-10 regulates early IL-12-mediated immune responses induced by the radiation-attenuated schistosome vaccine.
International immunology
Hogg KG,Kumkate S,Mountford AP
HM3500 was used in flow cytometry to investigate how IL-10 regulates IL-12 production in skin treated with radiation-attenuated schistosomes.
Mon Dec 01 00:00:00 EST 2003
CD8+ tumor-infiltrating lymphocytes are primed for Fas-mediated activation-induced cell death but are not apoptotic in situ.
Journal of immunology (Baltimore, Md. : 1950)
Radoja S,Saio M,Frey AB
HM3500 was used in flow cytometry to characterize murine tumor-infiltrating lymphocytes.
Tue May 15 00:00:00 EDT 2001
Inhibition of T lymphocyte activation in mice heterozygous for loss of the IMPDH II gene.
The Journal of clinical investigation
Gu JJ,Stegmann S,Gathy K,Murray R,Laliberte J,Ayscue L,Mitchell BS
HM3500 was used in blocking assay to characterize IMPDH II(-/-) and IMPDH II(+/-) mice.
Tue Aug 01 00:00:00 EDT 2000
Role of inflammatory infiltrate in activation and effector function of cloned islet reactive nonobese diabetic CD8+ T cells: involvement of a nitric oxide-dependent pathway.
Journal of immunology (Baltimore, Md. : 1950)
Gurlo T,Kawamura K,von Grafenstein H
HM3500 was used in blocking assay to study how CD8+ T cells interact with beta cells and local inflammatory cells in islets.
Wed Dec 01 00:00:00 EST 1999
CD8+ T cells become nonresponsive (anergic) following activation in the presence of costimulation.
Journal of immunology (Baltimore, Md. : 1950)
Deeths MJ,Kedl RM,Mescher MF
HM3500 was used in flow cytometry to study activation-induced nonresponsiveness of T cells.
Thu Jul 01 00:00:00 EDT 1999
Effects of dietary docosahexaenoic acid on surface molecules involved in T cell proliferation.
Biochimica et biophysica acta
Sasaki T,Kanke Y,Kudoh K,Misawa Y,Shimizu J,Takita T
HM3500 was used in flow cytometry to examine the effect of dietary docosahexaenoic acid on the surface molecules involved in T cell proliferation.
Mon Jan 04 00:00:00 EST 1999
Two types of anti-TL (thymus leukemia) CTL clones with distinct target specificities: differences in cytotoxic mechanisms and accessory molecule requirements.
Journal of immunology (Baltimore, Md. : 1950)
Tsujimura K,Takahashi T,Iwase S,Matsudaira Y,Kaneko Y,Yagita H,Obata Y
HM3500 was used in blocking assay to characterize TCRalphabeta CTL clones that recognize mouse thymus leukemia.
Mon Jun 01 00:00:00 EDT 1998
Targets of p56(lck) activity in immature thymoblasts: stimulation of the Ras/Raf/MAPK pathway.
International immunology
Lin K,Abraham KM
HM3500 was used in flow cytometry to demonstrate that the Ras/Raf/MAPK pathway is constitutively activated in lck-transformed immature thymoblasts.
Sat Feb 01 00:00:00 EST 1997