Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4ºC in the dark.
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
IGF1R (Insulin-like Growth Factor1 Receptor) is a receptor tyrosine kinase that mediates actions of insulin-like growth factor 1 (IFG1). It binds IGF1 with high affinity and IGF2/insulin with lower affinity. Activated IGF1R is involved in cell growth and survival control. IGF1R is crucial for tumor transformation and survial of malignant cells. Ligand binding activates the receptor kinase and leads to receptor autophosphorylation and tyrosines phosphorylation of multiple substrates that function as signaling adapter proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD221; IGF-I receptor; Insulin-like growth factor 1 receptor; Insulin-like growth factor I receptor; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Aliases: CD221; IGF1R; IGFIR; IGFR; JTK13
UniProt ID: (Human) P08069
Entrez Gene ID: (Human) 3480
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support