Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Membrane permeabilization is required for flow cytometry applications. For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4ºC in the dark. Once reconstituted, this product has a shelf life of approx. 18 months.
Mouse anti Human CD79a antibody, clone HM57 recognizes an epitope within the cytoplasmic domain of CD79a.
CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79a antigen (immunoglobulin-associated alpha); CD79a molecule, immunoglobulin-associated alpha; Ig alpha; Ig-alpha; immunoglobulin-associated alpha; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; Surface IgM-associated protein
Gene Aliases: CD79A; Ig-alpha; IGA; Igalpha; Ly-54; Ly54; MB-1; MB1
UniProt ID: (Human) P11912, (Mouse) P11911
Entrez Gene ID: (Human) 973, (Mouse) 12518, (Rat) 100913063
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support