Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • USP9X Antibodies

          Invitrogen

          USP9X Monoclonal Antibody (OTI2G7)

          Advanced Verification
          View all (35) USP9X antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite USP9X Monoclonal Antibody (OTI2G7)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (13)
          • Advanced Verification (1)
          USP9X Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          USP9X Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          USP9X Antibody (MA5-26367) in ICC/IF

          Immunofluorescence analysis of Probable ubiquitin carboxyl-terminal hydrolase FAF-X was performed using 70% confluent log phase A-431 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.01% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with USP9X Monoclonal Antibody (OTI2G7) (Product # MA5-26367) at 1:100 dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Goat anti-Mouse IgG (H+L) Superclonal™ Recombinan... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          USP9X Antibody in Immunocytochemistry (ICC/IF)
          USP9X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP9X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP9X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP9X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP9X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP9X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP9X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP9X Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          USP9X Antibody in Western Blot (WB)
          USP9X Antibody in Western Blot (WB)
          USP9X Antibody in Western Blot (WB)
          USP9X Antibody in Western Blot (WB)
          USP9X Antibody

          Product Details

          MA5-26367

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:150
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          OTI2G7

          Immunogen

          Human recombinant protein fragment corresponding to amino acids 2246-2570 of USP9X produced in E.coli

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 1% BSA, 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2723770

          Target Information

          USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity. USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Deubiquitinating enzyme FAF-X; Drosophila fat facets related, X-linked; Fat facets homolog; Fat facets in mammals; fat facets protein related, X-linked; Fat facets protein-related, X-linked; fat facets-like, X-linked; fats facets protein related, X; hFAM; probable ubiquitin carboxyl-terminal hydrolase FAF-X; similar to Homo sapiens ubiquitin hydrolase encoded by GenBank Accession Number X98296; Ubiquitin carboxyl-terminal hydrolase 9X; Ubiquitin carboxyl-terminal hydrolase FAM; ubiquitin specific peptidase 9, X chromosome; ubiquitin specific protease 9 SSSRF- isoform; ubiquitin specific protease 9, X chromosome; ubiquitin specific protease 9, X chromosome (fat facets-like Drosophila); Ubiquitin thioesterase FAF-X; ubiquitin thiolesterase FAF-X; ubiquitin-specific processing protease FAF-X; Ubiquitin-specific protease 9, X chromosome; Ubiquitin-specific-processing protease FAF-X; unnamed protein product

          View more View less

          Gene Aliases: 5730589N07Rik; AA407302; AA407699; AL022658; AL022749; DFFRX; FAF; FAF-X; Fafl; FAM; hFAM; MRX99; MRXS99F; RGD1560056; USP9; USP9X; XLID99

          View more View less

          UniProt ID: (Human) Q93008, (Mouse) P70398

          View more View less

          Entrez Gene ID: (Dog) 480885, (Human) 8239, (Mouse) 22284, (Rat) 363445

          View more View less

          Function(s)
          cysteine-type endopeptidase activity thiol-dependent ubiquitin-specific protease activity protein binding peptidase activity cysteine-type peptidase activity hydrolase activity Lys63-specific deubiquitinase activity co-SMAD binding ubiquitinyl hydrolase activity molecular sequestering activity K11-linked deubiquitinase activity Lys48-specific deubiquitinase activity thiol-dependent ubiquitinyl hydrolase activity cysteine protease
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter neuron migration DNA dealkylation involved in DNA repair proteolysis chromosome segregation transforming growth factor beta receptor signaling pathway female gamete generation protein localization cell migration protein import into peroxisome matrix protein import into peroxisome matrix, receptor recycling protein ubiquitination protein deubiquitination BMP signaling pathway regulation of protein stability positive regulation of protein binding negative regulation of proteasomal ubiquitin-dependent protein catabolic process monoubiquitinated protein deubiquitination regulation of circadian rhythm rhythmic process axon extension protein stabilization cell division cilium morphogenesis cytosolic ciliogenesis protein K63-linked deubiquitination protein deubiquitination involved in ubiquitin-dependent protein catabolic process negative regulation of cilium assembly positive regulation of TORC2 signaling amyloid fibril formation in utero embryonic development ubiquitin-dependent protein catabolic process cell cycle mitotic nuclear division post-embryonic development cerebellar cortex structural organization hippocampus development proteasome-mediated ubiquitin-dependent protein catabolic process regulation of proteasomal protein catabolic process cellular response to transforming growth factor beta stimulus neuron projection extension
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline