Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TPH2 Antibodies

          Invitrogen

          TPH2 Polyclonal Antibody

          Advanced Verification
          12 Published Figures
          18 References
          View all (13) TPH2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TPH2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (12)
          • Advanced Verification (1)
          TPH2 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          TPH2 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TPH2 Antibody (PA1-778) in IP

          Immunoprecipitation of TPH2 from raphe nuclei isolated from Sprague-Dawley rat brains using TPH2 Polyclonal Antibody (Product # PA1-778). Product # PA1-778 was coupled to magnetic beads and added to raphe nuclei whole cell extracts for 15 minutes at RT. Immunoprecipitated proteins were assessed by Western blotting (WB), using either mouse anti-TPH2 (Ms TPH2, Lane 2) or sheep anti-TPH2 antibodies (Sh TPH2, Lane 3). As a positive control for the Western blot, non-immunoprecipitated brain lysates were probed with the mouse anti-TPH2 antibody ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TPH2 Antibody in Immunoprecipitation (IP)
          TPH2 Antibody in Immunohistochemistry (IHC)
          TPH2 Antibody in Immunohistochemistry (IHC)
          TPH2 Antibody in Immunohistochemistry (IHC)
          TPH2 Antibody in Immunohistochemistry (IHC)
          TPH2 Antibody in Immunohistochemistry (IHC)
          TPH2 Antibody in Immunohistochemistry (IHC)
          TPH2 Antibody in Immunohistochemistry (IHC)
          TPH2 Antibody in Immunohistochemistry (IHC)
          TPH2 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          TPH2 Antibody in Western Blot (WB)
          TPH2 Antibody in Western Blot (WB)
          TPH2 Antibody in Western Blot (WB)
          TPH2 Antibody
          TPH2 Polyclonal Antibody

          Product Details

          PA1-778

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 5 publications 5 publications

          Immunohistochemistry (IHC)

          -
          View 9 publications 9 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:500
          -

          Immunoprecipitation (IP)

          1:1,000
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rabbit, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to the N-terminal region of TPH2: R(15) R G L S L D S A V P E D H Q L (30)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.1% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2207687

          Product Specific Information

          PA1-778 detects TPH2 in human, mouse, rat, rabbit and non-human primate samples.

          PA1-778 has been successfully used in immunocytochemistry, immunoprecipitation, and Western blot procedures. By Western blot PA1-778 detects a 56 kDa protein representing TPH2.

          The PA1-778 immunogen is a synthetic peptide corresponding to the N-terminal region of TPH2.

          Target Information

          Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: HSNTPH; MGC119994; Neuronal tryptophan hydroxylase; TPRH; TRPH; Tryptophan 5-hydroxylase 2; Tryptophan 5-monooxygenase 2; unnamed protein product

          View more View less

          Gene Aliases: ADHD7; AU043594; NTPH; TPH2

          View more View less

          UniProt ID: (Human) Q8IWU9, (Mouse) Q8CGV2, (Rat) Q8CGU9

          View more View less

          Entrez Gene ID: (Human) 121278, (Rabbit) 100344397, (Mouse) 216343, (Rat) 317675

          View more View less

          Function(s)
          monooxygenase activity tryptophan 5-monooxygenase activity iron ion binding oxidoreductase activity oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced pteridine as one donor, and incorporation of one atom of oxygen metal ion binding amino acid binding molecular_function
          Process(es)
          serotonin biosynthetic process from tryptophan metabolic process aromatic amino acid family metabolic process serotonin biosynthetic process oxidation-reduction process circadian rhythm response to activity response to nutrient levels response to estrogen response to glucocorticoid response to calcium ion cellular response to lithium ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline