Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Protein APC Antibodies

          Invitrogen

          Protein APC Polyclonal Antibody

          View all (40) Protein APC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Protein APC Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          Protein APC Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Protein APC Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Protein APC Antibody (PA5-119648) in ICC/IF

          Immunocytochemistry-Immunofluorescence analysis of APC in SH-SY-5Y cells (green) using Protein APC Polyclonal Antibody (Product # PA5-119648). The nuclear counter stain is DAPI (blue). Cells were fixed in paraformaldehyde, permeabilised with 0.25% Triton X100/PBS. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Protein APC Antibody in Immunocytochemistry (ICC/IF)
          Protein APC Antibody in Immunocytochemistry (ICC/IF)
          Protein APC Antibody in Immunocytochemistry (ICC/IF)
          Protein APC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Protein APC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Protein APC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Protein APC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Protein APC Antibody in Flow Cytometry (Flow)
          Protein APC Polyclonal Antibody

          Product Details

          PA5-119648

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1000
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide within C-terminal human Ifna1.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol, 0.2% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2913221

          Product Specific Information

          Positive Control: HUVEC, SH-SY-5Y, SiHa, rat brain tissue, human kidney tissue, human breast tissue, mouse colon tissue.

          Subcellular Location: Cell junction. Cytoplasm. Cytoskeleton. Membrane.

          Target Information

          This gene encodes a tumor suppressor protein that acts as an antagonist of the Wnt signaling pathway. It is also involved in other processes including cell migration and adhesion, transcriptional activation, and apoptosis. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli; adenomatous polyposis coli (APC); Adenomatous polyposis coli protein; APC; APC, WNT signaling pathway regulator; Deleted in polyposis 2.5; epididymis secretory sperm binding protein; FAP; FPC; multiple intestinal neoplasia; Protein APC; protein phosphatase 1, regulatory subunit 46; Unknown; unnamed protein product; WNT signaling pathway regulator

          View more View less

          Gene Aliases: AI047805; APC; AU020952; AW124434; BTPS2; CC1; DESMD; DP2; DP2.5; DP3; GS; mAPC; Min; PPP1R46; RATAPC

          View more View less

          UniProt ID: (Human) P25054, (Rat) P70478

          View more View less

          Entrez Gene ID: (Human) 324, (Mouse) 11789, (Rat) 24205

          View more View less

          Function(s)
          protein binding beta-catenin binding microtubule binding protein kinase regulator activity protein kinase binding ubiquitin protein ligase binding macromolecular complex binding gamma-catenin binding cadherin binding microtubule plus-end binding dynein complex binding protease binding protein complex binding
          Process(es)
          mitotic cytokinesis kidney development hair follicle development protein complex assembly cellular response to DNA damage stimulus negative regulation of microtubule depolymerization cell cycle arrest metaphase/anaphase transition of mitotic cell cycle mitotic spindle assembly checkpoint establishment or maintenance of cell polarity pattern specification process axonogenesis negative regulation of cell proliferation axis specification anterior/posterior pattern specification dorsal/ventral pattern formation proximal/distal pattern formation Wnt signaling pathway cell migration stem cell population maintenance negative regulation of Wnt signaling pathway regulation of cell migration positive regulation of cell migration regulation of epithelial cell differentiation positive regulation of epithelial cell differentiation positive regulation of microtubule polymerization cytoplasmic microtubule organization neuron projection development positive regulation of pseudopodium assembly regulation of microtubule-based process T cell differentiation in thymus somatic stem cell population maintenance negative regulation of odontogenesis positive regulation of apoptotic process negative regulation of apoptotic process negative regulation of MAPK cascade skin development canonical Wnt signaling pathway involved in negative regulation of apoptotic process canonical Wnt signaling pathway involved in positive regulation of apoptotic process regulation of cell differentiation positive regulation of cell differentiation regulation of osteoblast differentiation regulation of osteoclast differentiation positive regulation of protein catabolic process negative regulation of cyclin-dependent protein serine/threonine kinase activity positive regulation of cell adhesion muscle cell cellular homeostasis thymus development negative regulation of epithelial cell proliferation regulation of nitrogen compound metabolic process chromosome organization regulation of cell cycle positive regulation of cell division regulation of attachment of spindle microtubules to kinetochore retina development in camera-type eye canonical Wnt signaling pathway negative regulation of epithelial cell proliferation involved in prostate gland development negative regulation of canonical Wnt signaling pathway regulation of glutamate metabolic process cell fate specification heart valve development endocardial cushion morphogenesis cell adhesion nervous system development insulin receptor signaling pathway proteasome-mediated ubiquitin-dependent protein catabolic process regulation of microtubule-based movement protein-containing complex assembly bicellular tight junction assembly regulation of primary metabolic process positive regulation of cold-induced thermogenesis negative regulation of cell cycle G1/S phase transition positive regulation of protein localization to centrosome negative regulation of G1/S transition of mitotic cell cycle pancreas development response to drug response to alcohol cellular response to nerve growth factor stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline