Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00437306_m1
          See other VEGFA GE Assays ›
          Gene Symbol:
          Vegfa
          Gene Name:
          vascular endothelial growth factor A
          Gene Aliases:
          Vegf, Vpf
          Chromosome Location:
          Chr.17: 46016993 - 46032377 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00437306_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vegfa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
          RefSeq NM_001025250.3 NP_001020421.2 3 - 4 1328 61
          NM_001025257.3 NP_001020428.2 3 - 4 1328 61
          NM_001287056.1 NP_001273985.1 3 - 4 1328 61
          NM_001287057.1 NP_001273986.1 3 - 4 1328 61
          NM_001287058.1 NP_001273987.1 3 - 4 1328 61
          NM_009505.4 NP_033531.3 3 - 4 1328 61
          GenBank mRNA AY120866.1 - 1 - 2 206 61
          AY707864.1 - 4 - 5 525 61
          AY750956.1 - 4 - 5 486 61
          AY750957.1 - 4 - 5 449 61
          AY756068.1 - 3 - 4 264 61
          BC061468.1 - 3 - 4 701 61
          BQ832724.1 - 2 - 3 229 61
          EF028705.1 - 4 - 5 521 61
          M95200.1 - 3 - 4 393 61
          S37052.1 - 3 - 4 319 61
          S38083.1 - 3 - 4 319 61
          S38100.1 - 3 - 4 319 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002162.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vegfa

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          ovarian follicle development
          blood vessel development
          patterning of blood vessels
          response to hypoxia
          in utero embryonic development
          kidney development
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          vasculature development
          lymphangiogenesis
          blood vessel remodeling
          sprouting angiogenesis
          cell migration involved in sprouting angiogenesis
          positive regulation of neuroblast proliferation
          positive regulation of mesenchymal cell proliferation
          positive regulation of receptor internalization
          basophil chemotaxis
          heart morphogenesis
          outflow tract morphogenesis
          coronary vein morphogenesis
          regulation of transcription from RNA polymerase II promoter
          multicellular organism development
          nervous system development
          mesoderm development
          lactation
          cell proliferation
          positive regulation of cell proliferation
          regulation of cell shape
          positive regulation of signal transduction
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          cell migration
          negative regulation of cell-cell adhesion
          cell differentiation
          monocyte differentiation
          macrophage differentiation
          lung development
          positive regulation of cell migration
          regulation of cGMP metabolic process
          epithelial cell differentiation
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          post-embryonic camera-type eye development
          positive regulation of protein complex assembly
          positive regulation of protein autophosphorylation
          activation of protein kinase activity
          positive regulation of CREB transcription factor activity
          positive regulation of peptidyl-serine phosphorylation
          tube formation
          endothelial cell chemotaxis
          cellular response to vascular endothelial growth factor stimulus
          lymph vessel morphogenesis
          positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
          vascular endothelial growth factor signaling pathway
          positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
          VEGF-activated neuropilin signaling pathway
          growth
          T-helper 1 type immune response
          eye photoreceptor cell development
          negative regulation of apoptotic process
          negative regulation of programmed cell death
          positive regulation of vascular permeability
          surfactant homeostasis
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MAP kinase activity
          negative regulation of neuron apoptotic process
          positive regulation of blood vessel endothelial cell migration
          endothelial cell migration
          regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          negative regulation of bone resorption
          positive regulation of cell adhesion
          positive regulation of transcription from RNA polymerase II promoter
          vascular endothelial growth factor receptor signaling pathway
          mRNA stabilization
          lung alveolus development
          cell maturation
          blood vessel morphogenesis
          camera-type eye morphogenesis
          positive regulation of smooth muscle cell proliferation
          neuron development
          cardiac muscle fiber development
          branching morphogenesis of an epithelial tube
          positive regulation of axon extension involved in axon guidance
          artery morphogenesis
          positive regulation of epithelial cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          positive chemotaxis
          positive regulation of positive chemotaxis
          induction of positive chemotaxis
          positive regulation of cellular component movement
          positive regulation of cell division
          positive regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          primitive erythrocyte differentiation
          cell chemotaxis
          lung vasculature development
          mammary gland alveolus development
          positive regulation of mast cell chemotaxis
          cardiac vascular smooth muscle cell development
          angiogenesis involved in coronary vascular morphogenesis
          coronary artery morphogenesis
          positive regulation of transcription from RNA polymerase II promoter in response to hypoxia
          positive regulation of ERK1 and ERK2 cascade
          cellular response to hypoxia
          dopaminergic neuron differentiation
          commissural neuron axon guidance
          positive regulation of protein kinase C signaling
          positive regulation of cell migration involved in sprouting angiogenesis
          positive regulation of branching involved in ureteric bud morphogenesis
          regulation of retinal ganglion cell axon guidance
          positive regulation of peptidyl-tyrosine autophosphorylation
          positive regulation of p38MAPK cascade
          negative regulation of neuron death
          positive regulation of lymphangiogenesis
          regulation of hematopoietic progenitor cell differentiation
          positive regulation of histone deacetylase activity
          positive regulation of retinal ganglion cell axon guidance
          positive regulation of protein localization to early endosome
          positive regulation of protein kinase D signaling
          positive regulation of receptor activity
          negative regulation of extrinsic apoptotic signaling pathway
          fibronectin binding
          receptor binding
          cytokine activity
          platelet-derived growth factor receptor binding
          vascular endothelial growth factor receptor binding
          protein binding
          growth factor activity
          heparin binding
          neuropilin binding
          chemoattractant activity
          identical protein binding
          protein homodimerization activity
          vascular endothelial growth factor receptor 1 binding
          vascular endothelial growth factor receptor 2 binding
          protein heterodimerization activity
          receptor agonist activity
          extracellular matrix binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001025250.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001025250.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001025257.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001025257.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001110266.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001110266.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001110267.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001110267.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001110268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001110268.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001287056.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001287057.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_001287058.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_009505.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 NM_009505.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020297.mm.1 TC1700002162.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020295.mm.1 NM_001025250.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020295.mm.1 NM_001025250.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020295.mm.1 NM_001110266.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020295.mm.1 NM_001110266.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020295.mm.1 NM_001287056.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020295.mm.1 TC1700002162.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch