Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm00444911_m1
See other GSK3B GE Assays ›
Gene Symbol:
Gsk3b
Gene Name:
glycogen synthase kinase 3 beta
Gene Aliases:
7330414F15Rik, 8430431H08Rik, C86142, GSK-3, GSK-3beta, GSK3
Chromosome Location:
Chr.16: 38089001 - 38246080 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm00444911_m1
Best Coverage
Size
FAM (900 nM Primer) | XS: 75 rxns
FAM (900 nM Primer) | S: 250 rxns
FAM (900 nM Primer) | M: 750 rxns
FAM (900 nM Primer) | L: 2,900 rxns
FAM PL (350 nM Primer Limited) | S: 360 rxns
FAM PL (350 nM Primer Limited) | M: 750 rxns
FAM PL (350 nM Primer Limited) | L: 2,900 rxns
VIC (900 nM Primer) | S: 360 rxns
VIC (900 nM Primer) | M: 750 rxns
VIC (900 nM Primer) | L: 2,900 rxns
VIC PL (150 nM Primer Limited) | S: 360 rxns
VIC PL (150 nM Primer Limited) | M: 750 rxns
VIC PL (150 nM Primer Limited) | L: 2,900 rxns
ABY PL (350 nM Primer Limited) | S: 360 rxns
ABY PL (350 nM Primer Limited) | M: 750 rxns
ABY PL (350 nM Primer Limited) | L: 2,900 rxns
JUN PL (350 nM Primer Limited) | S: 360 rxns
JUN PL (350 nM Primer Limited) | M: 750 rxns
JUN PL (350 nM Primer Limited) | L: 2,900 rxns
CYN5 PL (350 nM Primer Limited) | S: 360 rxns
CYN5 PL (350 nM Primer Limited) | M: 750 rxns
CYN5 PL (350 nM Primer Limited) | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Citations (1)
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Gsk3b
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
Amplicon Length
RefSeq
NM_019827.6
NP_062801.1
8 - 9
2431
72
GenBank mRNA
AB066114.1
-
9 - 10
2430
72
AF156099.2
-
8 - 9
1072
72
AK154293.1
-
9 - 10
1909
72
AK170965.1
-
9 - 10
1878
72
BC006936.1
-
8 - 9
938
72
BC060743.1
-
9 - 10
2358
72
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC1600000542.mm.1
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Protein Kinase
Mammalian Gene Collection
Recommended Assay
Mus musculus
Mouse Wnt Panel
Mouse Alzheimer's Disease
1700
Inventoried
Mouse_Alzheimer
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Gsk3b
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
re-entry into mitotic cell cycle
epithelial to mesenchymal transition
positive regulation of cell-matrix adhesion
carbohydrate metabolic process
glycogen metabolic process
regulation of gene expression by genetic imprinting
protein phosphorylation
protein export from nucleus
ER overload response
cytoskeleton organization
establishment or maintenance of cell polarity
multicellular organism development
nervous system development
axonogenesis
myoblast fusion
circadian rhythm
cell proliferation
organ morphogenesis
negative regulation of signal transduction
negative regulation of cardiac muscle hypertrophy
positive regulation of peptidyl-threonine phosphorylation
regulation of neuron projection development
negative regulation of neuron projection development
negative regulation of neuron maturation
myotube differentiation
Wnt signaling pathway
phosphorylation
cell migration
peptidyl-serine phosphorylation
hippocampus development
establishment of cell polarity
cell differentiation
negative regulation of protein complex assembly
positive regulation of protein complex assembly
negative regulation of protein binding
positive regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of microtubule-based process
positive regulation of peptidyl-serine phosphorylation
protein localization to microtubule
intracellular signal transduction
cellular response to interleukin-3
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of MAP kinase activity
positive regulation of GTPase activity
hypermethylation of CpG island
canonical Wnt signaling pathway involved in positive regulation of apoptotic process
fat cell differentiation
positive regulation of axon extension
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
positive regulation of protein export from nucleus
regulation of neuronal synaptic plasticity
rhythmic process
negative regulation of dendrite morphogenesis
negative regulation of NFAT protein import into nucleus
canonical Wnt signaling pathway
superior temporal gyrus development
cellular response to mechanical stimulus
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
positive regulation of stem cell differentiation
nucleotide binding
RNA polymerase II transcription factor binding
p53 binding
protein kinase activity
protein serine/threonine kinase activity
integrin binding
protein binding
ATP binding
beta-catenin binding
transcription factor binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
ionotropic glutamate receptor binding
tau protein binding
tau-protein kinase activity
NF-kappaB binding
MicroArray Probes:
ABI
Array Name
Probe IDs
Targets
Target Type
ABI Mouse Genome Survey Microarray
564783
-
-
Back To Top
Citations (1)
Transgenic mice overexpressing glycogen synthase kinase 3beta: a putative model of hyperactivity and mania.
Authors
Prickaerts J, Moechars D, Cryns K, Lenaerts I, van Craenendonck H, Goris I, Daneels G, Bouwknecht JA, Steckler T
Journal
J Neurosci - Page No 9022 (Wed Aug 30 00:00:00 UTC 2006)
Volume (Year)
26 (2006)
Product Usage
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.60.1-2026-secondary-format-patch