Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm00487530_m1
See other SMAD2 GE Assays ›
Gene Symbol:
Smad2
Gene Name:
SMAD family member 2
Gene Aliases:
7120426M23Rik, Madh2, Madr2, Smad-2, mMad2
Chromosome Location:
Chr.18: 76241251 - 76311748 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm00487530_m1
Best Coverage
Size
FAM (900 nM Primer) | XS: 75 rxns
FAM (900 nM Primer) | S: 250 rxns
FAM (900 nM Primer) | M: 750 rxns
FAM (900 nM Primer) | L: 2,900 rxns
FAM PL (350 nM Primer Limited) | S: 360 rxns
FAM PL (350 nM Primer Limited) | M: 750 rxns
FAM PL (350 nM Primer Limited) | L: 2,900 rxns
VIC (900 nM Primer) | S: 360 rxns
VIC (900 nM Primer) | M: 750 rxns
VIC (900 nM Primer) | L: 2,900 rxns
VIC PL (150 nM Primer Limited) | S: 360 rxns
VIC PL (150 nM Primer Limited) | M: 750 rxns
VIC PL (150 nM Primer Limited) | L: 2,900 rxns
ABY PL (350 nM Primer Limited) | S: 360 rxns
ABY PL (350 nM Primer Limited) | M: 750 rxns
ABY PL (350 nM Primer Limited) | L: 2,900 rxns
JUN PL (350 nM Primer Limited) | S: 360 rxns
JUN PL (350 nM Primer Limited) | M: 750 rxns
JUN PL (350 nM Primer Limited) | L: 2,900 rxns
CYN5 PL (350 nM Primer Limited) | S: 360 rxns
CYN5 PL (350 nM Primer Limited) | M: 750 rxns
CYN5 PL (350 nM Primer Limited) | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Smad2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001252481.1
NP_001239410.1
6 - 7
1175
72
NM_010754.5
NP_034884.2
6 - 7
1064
72
GenBank mRNA
AF005743.1
-
5 - 6
792
72
AK007817.1
-
6 - 7
841
72
AY334552.1
-
4 - 5
643
72
BC021342.1
-
6 - 7
863
5066237
72
BC089184.1
-
6 - 7
1175
5322611
72
U60530.1
-
5 - 6
792
72
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC1800000816.mm.1
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Recommended Assay
Mus musculus
Mouse Tumor Metastasis Panel
1700
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Smad2
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
endoderm formation
mesoderm formation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein phosphorylation
transforming growth factor beta receptor signaling pathway
common-partner SMAD protein phosphorylation
SMAD protein complex assembly
zygotic specification of dorsal/ventral axis
gastrulation
pattern specification process
endoderm development
heart development
negative regulation of cell proliferation
response to glucose
post-embryonic development
embryonic pattern specification
anterior/posterior pattern specification
positive regulation of gene expression
negative regulation of gene expression
positive regulation of epithelial to mesenchymal transition
regulation of transforming growth factor beta receptor signaling pathway
signal transduction involved in regulation of gene expression
insulin secretion
lung development
positive regulation of BMP signaling pathway
pancreas development
activin receptor signaling pathway
organ growth
intracellular signal transduction
nodal signaling pathway
cell fate commitment
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
paraxial mesoderm morphogenesis
developmental growth
embryonic foregut morphogenesis
embryonic cranial skeleton morphogenesis
regulation of binding
palate development
pericardium development
SMAD protein signal transduction
response to cholesterol
positive regulation of nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transforming growth factor beta receptor binding
protein binding
transcription factor binding
phosphatase binding
transforming growth factor beta receptor, pathway-specific cytoplasmic mediator activity
ubiquitin protein ligase binding
activating transcription factor binding
type I transforming growth factor beta receptor binding
protein homodimerization activity
SMAD binding
metal ion binding
protein heterodimerization activity
co-SMAD binding
I-SMAD binding
R-SMAD binding
MicroArray Probes:
Affymetrix
ABI
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR1800005894.mm.1
NM_001252481.1.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1800005894.mm.1
NM_001252481.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1800005894.mm.1
NM_010754.5.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1800005894.mm.1
NM_010754.5
Refseq
Affymetrix Mouse Clariom S Array
PSR1800005894.mm.1
TC1800000816.mm.1
MTA TC
Array Name
Probe IDs
Targets
Target Type
ABI Mouse Genome Survey Microarray
367905
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-69bcbb9f7b-sgq2d:80/100.66.78.224:80.
git-commit: ba3ee22dad6ec1e333d6fdee729a43ee1272bbcb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.59.0-PolychromeNPI