Search
Search

MRH6| Gene Symbol: MRH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335200.1 | NP_001323815.1 | 1 - 2 | 359 | 117 | |
| NM_126419.1 | NP_178467.1 | 1 - 2 | 309 | 117 | ||
| GenBank mRNA | BT010857.1 | - | 1 - 2 | 205 | 117 | |
| BT020330.1 | - | 1 - 2 | 115 | 117 | ||
Set Membership: |
|