Search
Search

PDCB5| Gene Symbol: PDCB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339907.1 | NP_001327504.1 | 1 - 2 | 1765 | 98 | |
| NM_115672.3 | NP_567060.1 | 1 - 2 | 566 | 98 | ||
| GenBank mRNA | BT025769.1 | - | 1 - 2 | 355 | 98 | |
Set Membership: |
|