Search
Search

ATAUX2-11| Gene Symbol: ATAUX2-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123736.3 | NP_199183.1 | 1 - 2 | 466 | 153 | |
| GenBank mRNA | AK227549.1 | - | 1 - 2 | 239 | 153 | |
| BT005853.1 | - | 1 - 2 | 180 | 153 | ||
| L15450.1 | - | 1 - 2 | 180 | 153 | ||
Set Membership: |
|