Search
Search

MET1| Gene Symbol: MET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344823.1 | NP_001331134.1 | 5 - 6 | 3006 | 128 | |
| NM_124293.4 | NP_199727.1 | 4 - 5 | 3315 | 128 | ||
| GenBank mRNA | L10692.1 | - | 4 - 5 | 3208 | 128 | |
Set Membership: |
|