Search
Search

MYB3| Gene Symbol: MYB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332554.1 | NP_001322404.1 | 1 - 2 | 356 | 163 | |
| NM_102111.4 | NP_564176.2 | 1 - 2 | 356 | 163 | ||
| GenBank mRNA | AY072543.1 | - | 1 - 2 | 224 | 163 | |
| BT000841.1 | - | 1 - 2 | 131 | 163 | ||
Set Membership: |
|