Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Ba07922409_po
          Gene Symbol:
          Bacteria
          Gene Name:
          class A extended-spectrum beta-lactamase CTX-M (blaCTX-M)
          Gene Aliases:
          Bacteria, blaCTX-M-46, blaCTX-M-47, blaCTX-M-48, blaCTX-M-49, blaCTX-M-50, blaCTX-M-51, blaCTX-M-52, blaCTX-M-53, blaCTX-M-54, blaCTX-M-55/blaCTX-M-57, blaCTX-M-58, blaCTX-M-60, blaCTX-M-61, blaCTX-M-62, blaCTX-M-63, blaCTX-M-64, blaCTX-M-65, blaCTX-M-66, blaCTX-M-67, blaCTX-M-68, blaCTX-M-69, blaCTX-M-71, blaCTX-M-72, blaCTX-M-73, blaCTX-M-78, blaCTX-M-79, blaCTX-M-8, blaCTX-M-80, blaCTX-M-82, blaCTX-M-83, blaCTX-M-84, blaCTX-M-85, blaCTX-M-86, blaCTX-M-87, blaCTX-M-88, blaCTX-M-89, blaCTX-M-9, blaCTX-M-90, blaCTX-M-91, blaCTX-M-93, blaCTX-M-94, blaCTX-M-96, blaCTX-M-98, blaCTX-M-99, blaCTX-M-1, blaCTX-M-10, blaCTX-M-100, blaCTX-M-101, blaCTX-M-102, blaCTX-M-103, blaCTX-M-104, blaCTX-M-105, blaCTX-M-110, blaCTX-M-111, blaCTX-M-112, blaCTX-M-113, blaCTX-M-114, blaCTX-M-116, blaCTX-M-117, blaCTX-M-12, blaCTX-M-121, blaCTX-M-122, blaCTX-M-123, blaCTX-M-125, blaCTX-M-126, blaCTX-M-127, blaCTX-M-129, blaCTX-M-13, blaCTX-M-130, blaCTX-M-134, blaCTX-M-136, blaCTX-M-137, blaCTX-M-138, blaCTX-M-139, blaCTX-M-14/blaCTX-M-18, blaCTX-M-142, blaCTX-M-143, blaCTX-M-144, blaCTX-M-146, blaCTX-M-147, blaCTX-M-148, blaCTX-M-15/blaUOE-1, blaCTX-M-150, blaCTX-M-152, blaCTX-M-153, blaCTX-M-154, blaCTX-M-155, blaCTX-M-156, blaCTX-M-157, blaCTX-M-158, blaCTX-M-159, blaCTX-M-16, blaCTX-M-160, blaCTX-M-161, blaCTX-M-162, blaCTX-M-163, blaCTX-M-164, blaCTX-M-166, blaCTX-M-167, blaCTX-M-168, blaCTX-M-169, blaCTX-M-17/blaCTX-M-14b, blaCTX-M-170, blaCTX-M-172, blaCTX-M-173, blaCTX-M-174, blaCTX-M-176, blaCTX-M-177, blaCTX-M-178, blaCTX-M-179, blaCTX-M-180, blaCTX-M-181, blaCTX-M-182, blaCTX-M-183, blaCTX-M-184, blaCTX-M-185, blaCTX-M-186, blaCTX-M-187, blaCTX-M-188, blaCTX-M-189, blaCTX-M-19, blaCTX-M-190, blaCTX-M-191, blaCTX-M-192, blaCTX-M-193, blaCTX-M-194, blaCTX-M-195, blaCTX-M-196, blaCTX-M-197, blaCTX-M-198, blaCTX-M-199, blaCTX-M-201, blaCTX-M-202, blaCTX-M-203, blaCTX-M-204, blaCTX-M-205, blaCTX-M-206, blaCTX-M-207, blaCTX-M-208, blaCTX-M-209, blaCTX-M-21, blaCTX-M-210, blaCTX-M-211, blaCTX-M-212, blaCTX-M-213, blaCTX-M-214, blaCTX-M-215, blaCTX-M-216, blaCTX-M-217, blaCTX-M-218, blaCTX-M-219, blaCTX-M-22, blaCTX-M-220, blaCTX-M-221, blaCTX-M-222, blaCTX-M-223, blaCTX-M-225, blaCTX-M-226, blaCTX-M-227, blaCTX-M-228, blaCTX-M-23, blaCTX-M-230, blaCTX-M-231, blaCTX-M-232, blaCTX-M-233, blaCTX-M-235, blaCTX-M-236, blaCTX-M-237, blaCTX-M-238, blaCTX-M-239, blaCTX-M-24, blaCTX-M-240, blaCTX-M-241, blaCTX-M-242, blaCTX-M-243, blaCTX-M-244, blaCTX-M-245, blaCTX-M-25, blaCTX-M-251, blaCTX-M-252, blaCTX-M-26, blaCTX-M-27, blaCTX-M-28, blaCTX-M-29, blaCTX-M-3/blaCTX-M-133, blaCTX-M-30, blaCTX-M-32, blaCTX-M-33, blaCTX-M-34, blaCTX-M-36, blaCTX-M-37, blaCTX-M-38, blaCTX-M-39, blaCTX-M-40, blaCTX-M-41, blaCTX-M-42, class A extended-spectrum beta-lactamase CTX-M, blaCTX-M
          Chromosome Location:
          -
          Species:
          Bacteria
          Species Specific ID (Flybase ID):
          -
          Assay ID Ba07922409_po
          Best Coverage
          Size
          Availability Made To Order
          Catalog #
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details


          Back To Top

          More Information


          Updated Mapping Information

          This pooled assay is composed of the following assays: Ba04646149_s1, Ba04646154_s1, Ba04646127_s1.

          Set Membership:

          Recommended Assay Microbe Detection

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline