Search
Search

Ddx3x| Gene Symbol: Ddx3x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495366.2 | XP_003495414.1 | 9 - 10 | 975 | 70 | |
| XM_007640262.1 | XP_007638452.1 | 9 - 10 | 975 | 70 | ||
| XM_007640272.1 | XP_007638462.1 | 9 - 10 | 972 | 70 | ||
| GenBank mRNA | JI869484.1 | - | 5 - 6 | 587 | 70 | |
| JP064874.1 | - | 5 - 6 | 586 | 70 | ||
Set Membership: |
|