Search
Search

RpS4| Gene Symbol: RpS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300126.1 | NP_001287055.1 | 3 - 4 | 376 | 80 | |
| NM_079329.3 | NP_524053.2 | 2 - 3 | 475 | 80 | ||
| NM_168537.2 | NP_729871.1 | 3 - 4 | 376 | 80 | ||
| GenBank mRNA | BT011369.1 | - | 3 - 4 | 376 | 80 | |
Set Membership: |
|