Search
Search

Cyp4d2| Gene Symbol: Cyp4d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297877.1 | NP_001284806.1 | 1 - 2 | 1051 | 68 | |
| NM_080304.3 | NP_525043.1 | 1 - 2 | 166 | 68 | ||
| GenBank mRNA | AY118763.1 | - | 1 - 2 | 166 | 68 | |
Set Membership: |
|