Search
Search

wt1a| Gene Symbol: wt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131046.2 | NP_571121.1 | 3 - 4 | 948 | 83 | |
| GenBank mRNA | AF144550.1 | - | 3 - 4 | 697 | 83 | |
| AY028627.1 | - | 3 - 4 | 559 | 83 | ||
| BC162349.1 | - | 3 - 4 | 657 | 83 | ||
| BC162638.1 | - | 3 - 4 | 657 | 83 | ||
Set Membership: |
|