Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00153138_m1
          See other CCNA2 GE Assays ›
          Gene Symbol:
          CCNA2
          Gene Name:
          cyclin A2
          Gene Aliases:
          CCN1, CCNA
          Chromosome Location:
          Chr.4: 121816444 - 121823933 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00153138_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCNA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001237.3 NP_001228.1 1 - 2 520 110
          GenBank mRNA AK291931.1 - 1 - 2 503 110
          BC104783.1 - 1 - 2 315 8143786 110
          BC104787.1 - 1 - 2 315 8143790 110
          CR407692.1 - 1 - 2 215 110
          DA748568.1 - 1 - 2 520 110
          X51688.1 - 1 - 2 322 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400011748.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001516.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens HUMAN CYCLINS AND CELL CYCLE REGULATION HUMAN ENOS SIGNALING HUMAN BREAST CANCER REGULATION BY STATHMIN1 HUMAN PANCREATIC ADENOCARCINOMA Inventoried MicroArray Quality Control (MAQC) Human_Pancreatic_Adenocarcinoma Human_eNOS_Signaling Human_Cyclins_and_Cell_Cycle_Regulation Human_Breast_Cancer_Regulation_by_Stathmin1 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCNA2

          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          mitotic nuclear division
          mitotic G2 DNA damage checkpoint
          Ras protein signal transduction
          viral process
          organ regeneration
          response to glucagon
          cellular response to platelet-derived growth factor stimulus
          cellular response to leptin stimulus
          positive regulation of transcription, DNA-templated
          positive regulation of fibroblast proliferation
          cell division
          cellular response to cocaine
          cellular response to luteinizing hormone stimulus
          cellular response to estradiol stimulus
          cellular response to hypoxia
          cellular response to nitric oxide
          cochlea development
          cellular response to insulin-like growth factor stimulus
          protein binding
          protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04021536.hg.1 TC04001516.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04021536.hg.1 ENST00000274026.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04021536.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04021538.hg.1 TC04001516.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04021538.hg.1 ENST00000274026.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04021538.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04021537.hg.1 TC04001516.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04021537.hg.1 ENST00000274026.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04021537.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400180826.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180826.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180826.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400180826.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180826.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180826.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400180826.hg.1 TC0400011748.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400180827.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180827.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180827.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400180827.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180827.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180827.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400180827.hg.1 TC0400011748.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400180825.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180825.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180825.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400180825.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180825.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180825.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400180825.hg.1 TC0400011748.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400180828.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180828.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180828.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400180828.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180828.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400180828.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay PSR0400180828.hg.1 TC0400011748.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000274026.9 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 ENST00000618014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq
          Affymetrix Human Clariom D Assay TC0400011748.hg.1 NM_001237.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0