Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00163869_m1
          See other CA2 GE Assays ›
          Gene Symbol:
          CA2
          Gene Name:
          carbonic anhydrase 2
          Gene Aliases:
          CA-II, CAC, CAII, Car2, HEL-76, HEL-S-282
          Chromosome Location:
          Chr.8: 85463902 - 85481492 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00163869_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000067.2 NP_000058.1 2 - 3 416 85
          GenBank mRNA AK312978.1 - 2 - 3 311 85
          BC011949.1 - 2 - 3 301 3863603 85
          BC035424.1 - 1 - 2 206 85
          CR536526.1 - 2 - 3 236 85
          CR541875.1 - 2 - 3 236 85
          DA431650.1 - 2 - 3 423 85
          FJ224289.1 - 2 - 3 416 85
          GQ472235.1 - 2 - 3 301 85
          J03037.1 - 2 - 3 301 85
          M36532.1 - 2 - 3 280 85
          Y00339.1 - 2 - 3 274 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800008126.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000531.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CA2

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          morphogenesis of an epithelium
          one-carbon metabolic process
          response to pH
          response to zinc ion
          carbon dioxide transport
          bicarbonate transport
          positive regulation of synaptic transmission, GABAergic
          positive regulation of cellular pH reduction
          angiotensin-activated signaling pathway
          odontogenesis of dentin-containing tooth
          response to estrogen
          regulation of anion transport
          positive regulation of osteoclast differentiation
          positive regulation of bone resorption
          secretion
          response to steroid hormone
          regulation of intracellular pH
          cellular response to fluid shear stress
          positive regulation of dipeptide transmembrane transport
          regulation of chloride transport
          arylesterase activity
          carbonate dehydratase activity
          protein binding
          zinc ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08007207.hg.1 TC08000531.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08007207.hg.1 ENST00000285379.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08007207.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800155787.hg.1 TC0800008126.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000285379.9 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000518231.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520127.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000520996.5 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 ENST00000522742.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_000067.2 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Affymetrix Human Clariom D Assay TC0800008126.hg.1 NM_001293675.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 164835 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0