Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00170359_m1
          See other PLPP3 GE Assays ›
          Gene Symbol:
          PLPP3
          Gene Name:
          phospholipid phosphatase 3
          Gene Aliases:
          Dri42, LPP3, PAP2B, PPAP2B, VCIP
          Chromosome Location:
          Chr.1: 56494747 - 56579584 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00170359_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PLPP3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003713.4 NP_003704.3 1 - 2 709 65
          GenBank mRNA AB000889.1 - 1 - 2 147 65
          AF017786.1 - 2 - 3 515 65
          AF043329.1 - 1 - 2 434 65
          AF480883.1 - 2 - 3 521 65
          AK312439.1 - 1 - 2 667 65
          BC009196.2 - 1 - 2 512 3960223 65
          BT019589.1 - 1 - 2 141 65
          U79294.1 - 2 - 3 515 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100018443.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002701.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens HUMAN RHOA PATHWAY 1700 Inventoried Human_RhoA_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PLPP3

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          gastrulation with mouth forming second
          negative regulation of protein phosphorylation
          protein dephosphorylation
          lipid metabolic process
          phospholipid metabolic process
          germ cell migration
          regulation of Wnt signaling pathway
          sphingolipid biosynthetic process
          homotypic cell-cell adhesion
          canonical Wnt signaling pathway involved in positive regulation of endothelial cell migration
          canonical Wnt signaling pathway involved in positive regulation of cell-cell adhesion
          canonical Wnt signaling pathway involved in positive regulation of wound healing
          phospholipid dephosphorylation
          positive regulation of peptidyl-tyrosine phosphorylation
          protein stabilization
          positive regulation of sequence-specific DNA binding transcription factor activity
          Bergmann glial cell differentiation
          regulation of sphingolipid mediated signaling pathway
          phosphoprotein phosphatase activity
          integrin binding
          protein binding
          phosphatidate phosphatase activity
          sphingosine-1-phosphate phosphatase activity
          lipid phosphatase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01042370.hg.1 TC01002701.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01042370.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042370.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000459962.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000459962.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000459962.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000459962.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042369.hg.1 TC01002701.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01042369.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042369.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042366.hg.1 TC01002701.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01042366.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042366.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042367.hg.1 TC01002701.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01042367.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042367.hg.1 NM_003713.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042368.hg.1 TC01002701.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01042368.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042368.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242144.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242144.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242144.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242144.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242144.hg.1 TC0100018443.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100242142.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242142.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242142.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242142.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242142.hg.1 TC0100018443.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100242141.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242141.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242141.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242141.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242141.hg.1 TC0100018443.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100242143.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242143.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242143.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242143.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242143.hg.1 TC0100018443.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100242145.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242145.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242145.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100242145.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100242145.hg.1 TC0100018443.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000371250.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000459962.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000459962.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000459962.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000459962.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 ENST00000472957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018443.hg.1 NM_003713.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 133742 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline