Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00170953_m1
          See other S100A6 GE Assays ›
          Gene Symbol:
          S100A6
          Gene Name:
          S100 calcium binding protein A6
          Gene Aliases:
          2A9, 5B10, CABP, CACY, PRA
          Chromosome Location:
          Chr.1: 153534599 - 153536241 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00170953_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: S100A6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014624.3 NP_055439.1 1 - 2 295 94
          GenBank mRNA AK312088.1 - 1 - 2 45 94
          BC001431.2 - 1 - 2 100 3138609 94
          BC009017.1 - 1 - 2 276 4157026 94
          BF726732.1 - 1 - 2 295 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015864.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003264.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK312088.1,BC001431.2,BC009017.1,NM_014624.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene S100A6

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          axonogenesis
          ion transmembrane transport
          positive regulation of fibroblast proliferation
          calcium ion binding
          protein binding
          tropomyosin binding
          zinc ion binding
          ion transmembrane transporter activity
          protein homodimerization activity
          S100 protein binding
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01050233.hg.1 TC01003264.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050233.hg.1 ENST00000368719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050233.hg.1 ENST00000368720.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050233.hg.1 ENST00000496817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050233.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050231.hg.1 TC01003264.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050231.hg.1 ENST00000368719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050231.hg.1 ENST00000496817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050231.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000462951.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000462951.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000462951.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050232.hg.1 TC01003264.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050232.hg.1 ENST00000368719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050232.hg.1 ENST00000496817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050232.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050235.hg.1 TC01003264.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050235.hg.1 ENST00000368719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050235.hg.1 ENST00000368720.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050235.hg.1 ENST00000496817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050235.hg.1 NM_014624.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01050230.hg.1 TC01003264.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01050230.hg.1 ENST00000368719.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050230.hg.1 ENST00000496817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01050230.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219145.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219145.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219145.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219145.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219145.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219145.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219145.hg.1 TC0100015864.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100219146.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219146.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219146.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219146.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219146.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219146.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219146.hg.1 TC0100015864.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000462951.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000462951.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000462951.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015864.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 ENST00000462951.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 ENST00000462776.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 ENST00000462951.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219144.hg.1 TC0100015864.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100219147.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219147.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219147.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219147.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219147.hg.1 ENST00000368719.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219147.hg.1 ENST00000368720.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219147.hg.1 ENST00000496817.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100219147.hg.1 NM_014624.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100219147.hg.1 TC0100015864.hg.1 Clariom D TC

          Back To Top
           

          Citations (1)

          Human X-box binding protein-1 confers both estrogen independence and antiestrogen resistance in breast cancer cell lines.
          Authors
          Gomez BP, Riggins RB, Shajahan AN, Klimach U, Wang A, Crawford AC, Zhu Y, Zwart A, Wang M, Clarke R
          Journal
          Faseb J - Page No 4013 (Sat Dec 01 00:00:00 EST 2007)
          Volume (Year)
          21 (2007)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0