Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00172113_m1
          See other VDR GE Assays ›
          Gene Symbol:
          VDR
          Gene Name:
          vitamin D (1,25- dihydroxyvitamin D3) receptor
          Gene Aliases:
          NR1I1, PPP1R163
          Chromosome Location:
          Chr.12: 47841537 - 47905031 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00172113_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: VDR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000376.2 NP_000367.1 2 - 3 161 62
          NM_001017535.1 NP_001017535.1 3 - 4 283 62
          NM_001017536.1 NP_001017536.1 2 - 3 552 62
          GenBank mRNA AK312267.1 - 2 - 3 153 62
          BC060832.1 - 2 - 3 524 30343975 62
          DA721726.1 - 3 - 4 292 62
          J03258.1 - 2 - 3 116 62
          X67482.1 - 1 - 2 51 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010559.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001439.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK312267.1,BC060832.1,J03258.1,NM_000376.2,NM_001017535.1,X67482.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Immune Tolerance Network Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene VDR

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          skeletal system development
          transcription initiation from RNA polymerase II promoter
          calcium ion transport
          cellular calcium ion homeostasis
          signal transduction
          lactation
          negative regulation of cell proliferation
          positive regulation of gene expression
          negative regulation of keratinocyte proliferation
          positive regulation of vitamin D 24-hydroxylase activity
          bile acid signaling pathway
          steroid hormone mediated signaling pathway
          positive regulation of keratinocyte differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          decidualization
          intestinal absorption
          positive regulation of apoptotic process involved in mammary gland involution
          regulation of calcidiol 1-monooxygenase activity
          mammary gland branching involved in pregnancy
          vitamin D receptor signaling pathway
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          protein binding
          zinc ion binding
          calcitriol receptor activity
          lithocholic acid receptor activity
          sequence-specific DNA binding
          retinoid X receptor binding
          vitamin D response element binding
          calcitriol binding
          lithocholic acid binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000229022.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000395324.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000546653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000547065.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000548664.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000549336.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550314.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 ENST00000550325.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_000376.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017535.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010559.hg.1 NM_001017536.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline