Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00172455_m1
          See other LIG4 GE Assays ›
          Gene Symbol:
          LIG4
          Gene Name:
          DNA ligase 4
          Gene Aliases:
          LIG4S
          Chromosome Location:
          Chr.13: 108207442 - 108218368 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00172455_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LIG4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002312.3 NP_002303.2 1 - 2 240 64
          NM_206937.1 NP_996820.1 2 - 3 119 64
          XM_005254057.4 XP_005254114.1 2 - 3 254 64
          XM_006719951.3 XP_006720014.1 2 - 3 1959 64
          XM_011521091.2 XP_011519393.1 3 - 4 581 64
          XM_011521092.2 XP_011519394.1 2 - 3 157 64
          XM_017020569.1 XP_016876058.1 3 - 4 2222 64
          XM_017020570.1 XP_016876059.1 3 - 4 501 64
          GenBank mRNA BC037491.2 - 2 - 3 119 5259632 64
          X83441.1 - 1 - 2 240 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300009740.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000867.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC037491.2,NM_002312.3,NM_206937.1,X83441.1,XM_005254057.4,XM_006719951.3,XM_011521091.2,XM_011521092.2,XM_017020569.1,XM_017020570.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens HUMAN DNA REPAIR MECHANISM 1700 Inventoried Human_DNA_Repair_Mechanism Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LIG4

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          single strand break repair
          in utero embryonic development
          pro-B cell differentiation
          DNA ligation
          lagging strand elongation
          nucleotide-excision repair, DNA gap filling
          double-strand break repair
          double-strand break repair via nonhomologous end joining
          cell cycle
          central nervous system development
          cell proliferation
          response to X-ray
          response to gamma radiation
          T cell differentiation in thymus
          V(D)J recombination
          immunoglobulin V(D)J recombination
          T cell receptor V(D)J recombination
          somatic stem cell population maintenance
          negative regulation of neuron apoptotic process
          isotype switching
          positive regulation of fibroblast proliferation
          positive regulation of neurogenesis
          DNA ligation involved in DNA recombination
          DNA ligation involved in DNA repair
          chromosome organization
          cell division
          neuron apoptotic process
          cellular response to lithium ion
          DNA biosynthetic process
          establishment of integrated proviral latency
          double-strand break repair via classical nonhomologous end joining
          positive regulation of chromosome organization
          DNA binding
          DNA ligase activity
          DNA ligase (ATP) activity
          protein binding
          ATP binding
          protein C-terminus binding
          ligase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13009683.hg.1 ENST00000356922.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009683.hg.1 ENST00000405925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009683.hg.1 ENST00000442234.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009683.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009683.hg.1 NM_002312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009683.hg.1 NM_206937.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009683.hg.1 TC13000867.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR13009681.hg.1 ENST00000356922.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009681.hg.1 ENST00000405925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009681.hg.1 ENST00000442234.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009681.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009681.hg.1 NM_002312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009681.hg.1 NM_206937.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009681.hg.1 TC13000867.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR13009682.hg.1 ENST00000356922.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009682.hg.1 ENST00000405925.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009682.hg.1 ENST00000442234.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13009682.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009682.hg.1 NM_002312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009682.hg.1 NM_206937.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13009682.hg.1 TC13000867.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164992.hg.1 TC1300009740.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164993.hg.1 TC1300009740.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300164994.hg.1 TC1300009740.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000356922.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000405925.2 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000442234.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000611712.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 ENST00000614526.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_001098268.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_002312.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
          Affymetrix Human Clariom D Assay TC1300009740.hg.1 NM_206937.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 142541 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gl4ql:80/100.66.76.174:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0