Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00172470_m1
          See other MGMT GE Assays ›
          Gene Symbol:
          MGMT
          Gene Name:
          O-6-methylguanine-DNA methyltransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 129467184 - 129770983 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00172470_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MGMT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002412.4 NP_002403.2 3 - 4 403 92
          XM_005252682.2 XP_005252739.1 3 - 4 440 92
          XM_017016275.1 XP_016871764.1 2 - 3 226 92
          GenBank mRNA AK293593.1 - 3 - 4 373 92
          BC000824.1 - 3 - 4 331 3454207 92
          BI520029.1 - 3 - 4 385 92
          BT006714.1 - 2 - 3 278 92
          DC387686.1 - 3 - 4 397 92
          DC416206.1 - 3 - 4 403 92
          KC249995.1 - 2 - 3 312 92
          M29971.1 - 3 - 4 374 92
          M31767.1 - 2 - 3 300 92
          M60761.1 - 2 - 3 293 92
          X54228.1 - 2 - 3 318 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000009327.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000927.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MGMT

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA ligation
          DNA methylation
          DNA dealkylation involved in DNA repair
          response to toxic substance
          cellular response to oxidative stress
          response to drug
          negative regulation of apoptotic process
          regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to ethanol
          response to folic acid
          mammary gland epithelial cell differentiation
          cellular response to organic cyclic compound
          cellular response to ionizing radiation
          positive regulation of double-strand break repair
          DNA binding
          damaged DNA binding
          methylated-DNA-[protein]-cysteine S-methyltransferase activity
          calcium ion binding
          methyltransferase activity
          DNA-methyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000306010.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000462672.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000462672.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000462672.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000462672.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000482547.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000482547.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000482547.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000482653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000482653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000482653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 ENST00000482653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 NM_002412.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 NM_002412.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 NM_002412.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 NM_002412.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 NM_002412.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 NM_002412.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 NM_002412.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 NM_002412.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000009327.hg.1 NM_002412.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0