Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00176119_m1
          See other AK1 GE Assays ›
          Gene Symbol:
          AK1 ST6GALNAC4-ST6GALNAC6-AK1
          Gene Name:
          adenylate kinase 1
          ST6GALNAC4-ST6GALNAC6-AK1 readthrough
          Gene Aliases:
          ADK, Adk1, CNSHA3, HTL-S-58j
          -
          Chromosome Location:
          Chr.9: 127866480 - 127879621 on Build GRCh38
          Chromosome Location:
          Chr.9: 127866486 - 127917041 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00176119_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000476.3 NP_000467.1 5 - 6 408 66
          NM_001318121.1 NP_001305050.1 5 - 6 463 66
          NM_001318122.2 NP_001305051.1 4 - 5 403 66
          GenBank mRNA AB021871.1 - 3 - 4 323 66
          AK223501.1 - 4 - 5 351 66
          AK313758.1 - 5 - 6 407 66
          BC001116.2 - 5 - 6 394 66
          BC100022.1 - 5 - 6 423 66
          BG772183.1 - 4 - 5 471 66
          BT019580.1 - 3 - 4 323 66
          CR541977.1 - 3 - 4 323 66
          CR542004.1 - 3 - 4 323 66
          CR975542.1 - 5 - 6 463 66
          DA077967.1 - 5 - 6 477 66
          EU794628.1 - 5 - 6 476 66
          GQ891290.1 - 3 - 4 323 66
          Gene Symbol: ST6GALNAC4-ST6GALNAC6-AK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_174625.1 13 - 14 3642 66
          NR_174627.1 11 - 12 3522 66
          NR_174628.1 11 - 12 2900 66
          NR_174629.1 10 - 11 2845 66
          NR_174630.1 11 - 12 2881 66
          NR_174631.1 10 - 11 2826 66
          NR_174632.1 10 - 11 2915 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900012277.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09002920.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Human Cholera Infection Inventoried Human_Cholera_Infection Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AK1
          Click here to find siRNAs for Gene ST6GALNAC4-ST6GALNAC6-AK1

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          ADP biosynthetic process
          nucleoside triphosphate biosynthetic process
          nucleobase-containing small molecule interconversion
          AMP metabolic process
          ATP metabolic process
          nucleoside monophosphate phosphorylation
          nucleotide binding
          adenylate kinase activity
          nucleoside diphosphate kinase activity
          ATP binding
          kinase activity
          transferase activity
          nucleobase-containing compound kinase activity
          dAMP kinase activity
          nucleoside phosphate kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09020284.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09020284.hg.1 ENST00000373156.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09020284.hg.1 ENST00000373176.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09020284.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09020284.hg.1 TC09002920.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900186711.hg.1 TC0900012277.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000223836.10 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373156.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000373176.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000413016.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550143.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 ENST00000550992.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq
          Affymetrix Human Clariom D Assay TC0900012277.hg.1 NM_000476.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline