Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00181213_m1
          See other IGFBP5 GE Assays ›
          Gene Symbol:
          IGFBP5
          Gene Name:
          insulin like growth factor binding protein 5
          Gene Aliases:
          IBP5
          Chromosome Location:
          Chr.2: 216672105 - 216695549 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00181213_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IGFBP5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000599.3 NP_000590.1 1 - 2 1110 85
          GenBank mRNA AF055033.1 - 1 - 2 1087 85
          AK314992.1 - 1 - 2 388 85
          BC011453.1 - 1 - 2 684 4186607 85
          BT019706.1 - 1 - 2 336 85
          BT019707.1 - 1 - 2 336 85
          CR456809.1 - 1 - 2 336 85
          M62782.1 - 1 - 2 392 85
          M65062.1 - 1 - 2 379 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200015678.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002758.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Androgen Panel 1700 Inventoried MicroArray Quality Control (MAQC) Human_Androgen Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IGFBP5

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          osteoblast differentiation
          glucose metabolic process
          signal transduction
          female pregnancy
          regulation of glucose metabolic process
          negative regulation of smooth muscle cell migration
          negative regulation of translation
          negative regulation of cell migration
          hair follicle morphogenesis
          intracellular signal transduction
          glucose homeostasis
          positive regulation of insulin-like growth factor receptor signaling pathway
          negative regulation of insulin-like growth factor receptor signaling pathway
          cellular protein metabolic process
          type B pancreatic cell proliferation
          negative regulation of osteoblast differentiation
          negative regulation of growth
          lung alveolus development
          skeletal muscle tissue growth
          negative regulation of smooth muscle cell proliferation
          striated muscle cell differentiation
          positive regulation of protein kinase B signaling
          mammary gland involution
          response to growth hormone
          cellular response to cAMP
          cellular response to organic cyclic compound
          negative regulation of muscle tissue development
          negative regulation of skeletal muscle hypertrophy
          fibronectin binding
          protein binding
          insulin-like growth factor I binding
          insulin-like growth factor II binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02044326.hg.1 TC02002758.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02044326.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044326.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000447289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000447289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000447289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000447289.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000564299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000564299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000607591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000607591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000607591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010748.hg.1 ENST00000607591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044327.hg.1 TC02002758.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02044327.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044327.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02044325.hg.1 TC02002758.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02044325.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044325.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02044323.hg.1 TC02002758.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02044323.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044323.hg.1 NM_000599.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02044328.hg.1 TC02002758.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02044328.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02044328.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215100.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215100.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215100.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215100.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215100.hg.1 TC0200015678.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200215095.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215095.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215095.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215095.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215095.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215095.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215095.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215095.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215095.hg.1 TC0200015678.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200215101.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215101.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215101.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215101.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215101.hg.1 TC0200015678.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000447289.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000447289.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000447289.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000447289.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000564299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000564299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000607591.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000607591.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000607591.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200010748.hg.1 ENST00000607591.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215097.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215097.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215097.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215097.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215097.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215097.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215097.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215097.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215097.hg.1 TC0200015678.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 ENST00000486341.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay TC0200015678.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215098.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215098.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215098.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215098.hg.1 ENST00000233813.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215098.hg.1 ENST00000449583.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200215098.hg.1 NM_000599.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200215098.hg.1 TC0200015678.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 170211 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline